Applied Biosystems™

TaqMan™ SARS-CoV-2 Control Dilution Buffer

Catalog number: A49889
Have Questions?
Applied Biosystems™

TaqMan™ SARS-CoV-2 Control Dilution Buffer

Catalog number: A49889
Have Questions?
Catalog Number
A49889
Unit Size
10 x 250 µL
Price (USD)
Catalog NumberUnit SizePrice (USD)
A4988910 x 250 µLRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems TaqMan SARS-CoV-2 Control Dilution Buffer is used to dilute the TaqMan SARS-CoV-2 RNA Control (Cat. No. 956118), which is used with the TaqMan SARS-CoV-2 Pooling Assay Kit (Cat. No. A50790). This Dilution Buffer is also used to dilute the TaqMan SARS-CoV-2, Flu A, Flu B RNA Control and TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control, which are used with the TaqMan SARS-CoV-2, Flu A, Flu B RT-PCR Assay Kit (Cat. No. A47701) and TaqMan SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit (Cat. No. A47702).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-Probe Detection
For Use With (Application)
SARS CoV-2 Pathogen Research Solutions
Product Line
TaqMan
Quantity
10 x 250 μL
Shipping Condition
Dry Ice
Species
Human
Target Organism Class
severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)

Contents & Storage

TaqMan SARS CoV-2 Control Dilution Buffer, 1 x 250 μL/kit, 10 kits included

Store at -10 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text