ViralSEQ™ Lentivirus Physical Titer Kits
ViralSEQ™ Lentivirus Physical Titer Kits
Applied Biosystems™

ViralSEQ™ Lentivirus Physical Titer Kits

A one-step RT-qPCR assay for the quantitation of genome-containing lentiviral particles in supernatants from cell-based bioproduction systems. Lentiviral quantitation is a critical quality attribute (CQA) in viral vector manufacturing for cell and gene-modified cell therapy development.
Have Questions?
Modifier l'affichagebuttonViewtableView
RéférenceComprend
A52597Lentivirus Physical Titer Kit
A52598Lentivirus Physical Titer Kit, Nucleic Acid Sample Prep Kit
Référence A52597
Prix (EUR)
2 710,00
Each
-
Ajouter au panier
Comprend:
Lentivirus Physical Titer Kit
Recurring order eligible. Learn more »
Prix (EUR)
2 710,00
Each
Ajouter au panier
Ask our AI about this Product
The Applied Biosystems ViralSEQ Lentivirus Physical Titer Kit is a one-step RT-qPCR assay for the quantitation of genome-containing lentiviral particles in supernatants from cell-based bioproduction systems. Lentiviral quantitation is a critical quality attribute (CQA) in viral vector manufacturing for cell and gene-modified cell therapy development.
ViralSEQ Lentivirus Physical Titer Kit is an RT-qPCR assay for the quantitation of genome-containing lentiviral particles in supernatants from cell-based bioproduction systems. The assay targets a conserved region (LTR) in the lentiviral vector genome and can be used with most lentiviral transfer plasmids. Compared to p24 ELISA methods, lentiviral quantitation by RT-qPCR is more reproducible and specific to genome-containing particles.

Features of the ViralSEQ Lentivirus Physical Titer Kit:
• Lentiviral genome specific RT-qPCR assay for accurate viral particle count
• Assay design compatible with more than 200 lentivirus transfer plasmids
• Includes master mix, TaqMan assay mix and RNA control
• Configuration with PrepSEQ kit available for optimized RNA recovery

Integrated workflow solution
The ViralSEQ Lentivirus Physical Titer Kit is part of an integrated workflow for viral vector characterization during biopharmaceutical manufacturing. The kit is compatible with the PrepSEQ Nucleic Acid Sample Preparation Kit (Cat. No. A50485), which offers both a manual and automated sample preparation workflow. For real-time PCR, the ViralSEQ kit has been validated on the Applied Biosystems 7500 Fast Real-Time PCR System and QuantStudio 5 Real-Time PCR System. Data analysis is streamlined using AccuSEQ Real-Time PCR Detection Software that provides accurate quantitation and security, audit, and e-signature capabilities, to help enable 21 CFR Pt 11 compliance.

Broad dynamic range
The kit includes a synthetic RNA control to generate a standard curve, as well as internal positive controls. It enables quantitation over 7 logs of dynamic range from 50 copies to 109 copies for a range of titer values. TaqMan Assay-based detection allows for highly specific quantitation.

Genomic detection and quantitation of viral particles
Achieve specific quantitation of genome-containing lentiviral particles using an RT-qPCR assay. Methods relying on measurement of HIV-1 p24 could overestimate viral particle count to include partial capsids and free p24. The ViralSEQ Lentivirus Physical Titer kit measures viral particles based on viral RNA, thereby improving quantitation specificity of genome-containing particles.

Complete lentivirus characterization
ViralSEQ Lentivirus Physical Titer Kit can be used with the ViralSEQ™ Lentivirus Proviral DNA Titer Kit (Cat. No. A53561) for seamless correlation of qPCR data post-harvest (genome-containing particle count) and post-transduction (vector copy number analysis and infectious particle count). These kits provide a convenient method to correlate total and infectious lentiviral particles, facilitating lentiviral vector analytics in process development, optimization, and manufacturing QC.

For Research Use Only. Not for use in diagnostic procedures. TaqMan is a trademark of Roche Molecular Systems, Inc., used under permission and license.

Spécifications
À utiliser avec (équipement)Real-time PCR instruments (QuantStudio5 or 7500 Fast)
ComprendLentivirus Physical Titer Kit
Nbre de réactions100 réactions
Quantité100 reactions
CibleLentivirus genomic target
Méthode de détectionAmorce-sonde
À utiliser avec (application)Virus Detection
Unit SizeEach
Contenu et stockage
qPCR reagents, RNA control
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documentation et téléchargements

Certificats

Numéro de lotCertificate TypeDateCatalog Number(s)
2412016Certificate of Analysis27 mai 2025A52597
2412017Certificate of Analysis27 mai 2025A52597
2407015Certificate of Analysis05 août 2024A52597
2401013Certificate of Analysis11 mars 2024A52597
2305011Certificate of Analysis13 juin 2023A52597
5 résultats affichés, recherchez un certificat spécifique ci-dessus

Safety Data Sheets

Customers who viewed this item also viewed



Partager la référence catalogue, le nom ou le lien.

1x1 image pixel for data collection