Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
Description | AQUAfast chlorine (total) reagent powder packs, 100 tests |
Type | Chlorine (total) |
EPA Approved Methods | Drinking Water: Standard Methods 4500-Cl G / ATP EPA-approved Orion Method AC4P72; Wastewater: Standard Methods 4500-Cl G-2000 / ATP EPA-approved Orion Method AC4P72 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Chlorine Total-DPD Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.02 to 2 mg/L Cl2 |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast ammonia reagent powder packs, 100 tests |
Type | Ammonia |
EPA Approved Methods | Wastewater: Standard Methods 4500-NH3 F-1997 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | Set of 2: 200 Ammonia Salicylate Powder Packs, 200 Ammonia Cyanurate Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.01 to 0.8 mg/L N |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Salicylate |
Unit Size | Each |
Description | AQUAfast copper (free) reagent powder packs, 100 tests |
Type | Copper (free) |
EPA Approved Methods | Wastewater: Method 8506 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Cu 1 Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.05 to 5 mg/L Cu |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Bicinchoninate |
Unit Size | Each |
Description | AQUAfast molybdate reagent powder packs, 100 tests |
Type | Molybdate/Molybdenum |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | Set of 3: 100 Molybdenum HR1 Powder Packs, 100 Molybdenum HR2 Powder Packs, 100 Molybdenum HR3 Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.5 to 66 mg/L MoO4 |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Mercaptoacetic Acid |
Unit Size | Each |
Description | AQUAfast nitrite reagent powder packs, 100 tests |
Type | Nitrite |
EPA Approved Methods | Drinking Water: ASTM D3867-90 (B) / Standard Methods 4500-NO2- B; Wastewater: Standard Methods 4500-NO2- B |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Nitri 3 Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.01 to 0.3 mg/L N |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Diazotization (Azo) |
Unit Size | Each |
Description | AQUAfast manganese HR reagent powder packs, 100 tests |
Type | Manganese |
EPA Approved Methods | Wastewater: Method 8034 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | Set of 2: 100 Manganese Citrate Buffer Powder Packs, 100 Sodium Periodate Powder Pack |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.1 to 18 mg/L Mn |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Periodate Oxidation |
Unit Size | Each |
Description | AQUAfast silica (high range) reagent powder packs, 100 tests |
Type | Silica |
EPA Approved Methods | Drinking Water: Standard Methods 4500-SiO2 C; Wastewater: Standard Methods 4500-SiO2 C |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | Set of 3: 100 Silica HR Acid Reagent Powder Packs, 100 Silica Citric Acid Powder Packs, 100 Silica HR Molybdate Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 1 to 90 mg/L SiO2 |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | Silicomolybdate |
Unit Size | Each |
Description | AQUAfast chlorine (free) reagent powder packs, 100 tests |
Type | Chlorine (free) |
EPA Approved Methods | Drinking Water: Standard Methods 4500-Cl G / ATP EPA-approved Orion Method AC4P71; Wastewater: Standard Methods 4500-Cl G-2000 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Chlorine Free-DPD Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.02 to 2 mg/L Cl2 |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | DPD |
Unit Size | Case of 100 |
Description | AQUAfast iron (ferro) reagent powder packs, 100 tests |
Type | Iron (ferro) |
EPA Approved Methods | Wastewater: Standard Methods 3500-Fe-1997 |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Ferro Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.02 to 3 mg/L Fe |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | 1,10-Phenanthroline |
Unit Size | Each |
Description | AQUAfast iron (total) reagent powder packs, 100 tests |
Type | Iron (total) |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter |
Includes | 100 Iron TPTZ Powder Packs |
Packaging | Powder Pack |
Quantity | 100 tests |
Range | 0.02 to 1.8 mg/L Fe |
Recommended Accessories | AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Test Method | TPTZ |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Test Method | Range | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AC4P72 | Each | DPD | 0.02 to 2 mg/L Cl2 | 32.85 Online price 36.50 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P12 | Each | Salicylate | 0.01 to 0.8 mg/L N | 213.30 Online price 237.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P29 | Each | Bicinchoninate | 0.05 to 5 mg/L Cu | 60.30 Online price 67.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P42 | Each | Mercaptoacetic Acid | 0.5 to 66 mg/L MoO4 | 138.60 Online price 154.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P46 | Each | Diazotization (Azo) | 0.01 to 0.3 mg/L N | 56.52 Online price 62.80 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P55 | Each | Periodate Oxidation | 0.1 to 18 mg/L Mn | 82.08 Online price 91.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P60 | Each | Silicomolybdate | 1 to 90 mg/L SiO2 | 99.00 Online price 110.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P71 | Case of 100 | DPD | 0.02 to 2 mg/L Cl2 | 32.85 Online price 36.50 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P78 | Each | 1,10-Phenanthroline | 0.02 to 3 mg/L Fe | 33.57 Online price 37.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4P79 | Each | TPTZ | 0.02 to 1.8 mg/L Fe | 45.90 Online price 51.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|