Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Orion™ Powder Chemistry Packs
Thermo Scientific™

Orion™ Powder Chemistry Packs

Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeRange
AC4P42Molybdate/Molybdenum0.5 to 66 mg/L MoO4
AC4P12Ammonia0.01 to 0.8 mg/L N
AC4P29Copper (free)0.05 to 5 mg/L Cu
AC4P46Nitrite0.01 to 0.3 mg/L N
AC4P55Manganese0.1 to 18 mg/L Mn
AC4P60Silica1 to 90 mg/L SiO2
AC4P71Chlorine (free)0.02 to 2 mg/L Cl2
AC4P72Chlorine (total)0.02 to 2 mg/L Cl2
AC4P78Iron (ferro)0.02 to 3 mg/L Fe
AC4P79Iron (total)0.02 to 1.8 mg/L Fe
AC4P82Sulfate5 to 100 mg/L SO4
AC4P95Phosphate (ortho)0.06 to 2.5 mg/L PO4
Catalog number AC4P42
Price (USD)
151.20
Online Exclusive
168.00
Save 16.80 (10%)
Each
Add to cart
Type:
Molybdate/Molybdenum
Range:
0.5 to 66 mg/L MoO4
Recurring order eligible. Learn more »
Price (USD)
151.20
Online Exclusive
168.00
Save 16.80 (10%)
Each
Add to cart
Ask our AI about this Product

Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.

  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast™ Colorimeters

Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter
IncludesSet of 3: 100 Molybdenum HR1 Powder Packs, 100 Molybdenum HR2 Powder Packs, 100 Molybdenum HR3 Powder Packs
PackagingPowder Pack
Quantity100 tests
Range0.5 to 66 mg/L MoO4
Recommended AccessoriesAQUAfast 24 mm round vials (Cat. No. AC2V24)
Test MethodMercaptoacetic Acid
TypeMolybdate/Molybdenum
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
ZB2H1228Certificate of AnalysisJun 17, 2025AC4P71
ZB0F1028Certificate of AnalysisJun 17, 2025AC4P71
ZA6E0628Certificate of AnalysisJun 16, 2025AC4P71
ZA1B0128Certificate of AnalysisJun 16, 2025AC4P71
AB2B1229Certificate of AnalysisMay 26, 2025AC4P71
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection