mirVana™ miRNA Isolation Kit, with phenol
<i>mir</i>Vana&trade; miRNA Isolation Kit, with phenol
Invitrogen™

mirVana™ miRNA Isolation Kit, with phenol

The mirVana miRNA Isolation Kit uses a rapid procedure to isolate small RNAs from tissue and cells using an efficientRead more
Have Questions?
Catalog NumberQuantity
AM156040 Preps
Catalog number AM1560
Price (USD)
483.00
Each
-
Add to cart
Quantity:
40 Preps
Recurring order eligible. Learn more »
Price (USD)
483.00
Each
Add to cart
Ask our AI about this Product
The mirVana miRNA Isolation Kit uses a rapid procedure to isolate small RNAs from tissue and cells using an efficient glass fiber filter (GFF)-based method. The method isolates total RNA ranging in size from kilobases down to 10-mers. Each kit contains sufficient reagents and consumables for either 40 isolations of total RNA (including small RNAs), or 20 separate large and small RNA fractions.

Features of this kit include:

• Efficient isolation of small RNA-containing total RNA
• Enrich for small RNA (less than 200 nt) to increase sensitivity in downstream analyses
• Simple 30 minute procedure
• Ideal for miRNA, siRNA, shRNA, and snRNA analysis
• Compatible with virtually all cell and tissue types

Efficient recovery of small RNA
Current RNA purification procedures rely on organic extraction, followed by alcohol precipitation or adsorption of nucleic acid molecules on a GFF or silicate matrix. Unfortunately, the former procedure requires alcohol precipitation, which is time consuming and inefficiently recovers small RNAs. The latter procedure typically results in the loss of substantial amounts of small RNA. The mirVana miRNA Isolation Kit uses organic extraction followed by purification on a GFF under specialized binding and wash conditions. As a result, the kit effectively recovers all RNA—from large mRNA and ribosomal RNA down to 10-mers—from virtually all cell and tissue types. The mirVana miRNA Isolation Kit enables the isolation of small RNA-containing total RNA from samples consisting of 102–107 cultured cells or 0.5–250 mg tissue.

Enrich for small RNA
Although the mirVana miRNA Isolation Kit efficiently purifies all RNA larger than 10 nt, it also includes procedures for isolating RNA fractions, specifically enriched or depleted in small RNA species. Through the enrichment procedure included in the mirVana miRNA Isolation Kit, RNA molecules of ∼200 nt and less can be efficiently purified away from the larger RNA species. The resulting RNA preparation is highly enriched for miRNAs, siRNAs, and/or snRNAs. Small RNA enrichment using the mirVana miRNA Isolation Kit procedure allows more sensitive small RNA detection with less background as compared to the same assay used with total RNA.

Accessory product
The mirVana miRNA Isolation Kit without Phenol is available for customers concerned about the cost of shipping organic chemicals such as Acid Phenol:Chloroform, which is included in the standard kit.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume100 μL
Final Product TypeSmall RNA
For Use With (Application)qRT-PCR, microarray analysis, Northern blotting
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Purification Time30 min. (After sample homogenization)
Quantity40 Preps
Shipping ConditionRoom Temperature
Starting Material AmountBacteria: ≤107 cells
Cells: ≤107
Plant: ≤250 mg
Tissue: ≤250 mg
Virus: ≤107 cells
Yeast: ≤107 cells
Enzymatic Reactions: varies
YieldCells: ≤16 μg RNA per 50,000 cells
Tissue: ≤160 μg per 5 mg
Isolation TechnologySpin Column (Glass Fiber Filter), Organic Extraction
Sample TypeBacteria, Cells, Plant, Tissue, Viruses, Yeast, Enzymatic Reactions
Unit SizeEach
Contents & Storage
• 30 mL miRNA Wash Solution I (room temperature)
• 50 mL Wash Solution 2/3 (room temperature)
• 80 Collection Tubes (room temperature)
• 40 Filter Cartridges (room temperature)
• 100 mL Lysis/Binding Buffer (4°C)
• 10 mL miRNA Homogenate Additive (4°C)
• 100 mL Acid-Phenol:Chloroform (4°C)
• 1.4 mL Gel Loading Buffer II (-20°C)
• 5 mL Elution Solution (-20°C, 4°C, or room temperature)
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What kit would you recommend for isolation of miRNA from tissue, cells, or liquid samples?

For tissue or cells, we recommend the mirVana miRNA Isolation Kit (Cat. No. AM1560), while for liquid samples, we recommend the mirVana Paris RNA and Native Protein Purification Kit (Cat. No. AM1556). For high-throughput miRNA isolation we recommend using the MagMAX mirVana Total RNA Isolation kit (Cat. No. A27828)

What are different methods for cleaning up my miRNA sample?

- Precipitation of miRNA: Follow the instructions for the mirVANA miRNA Isolation Kit or the mirVANA PARIS RNA and Native Protein Purification Kit.
- Double phenol/guanidium (chaotropic reagent) extraction: Disrupt tissue with phenol/guanidine, then extract with phenol, centrifuge, remove the supernatant, and precipitate the RNA.
- Glass-fiber filter: Disrupt the tissue in a chaotropic agent, bind the RNA to a glass-fiber filter, then wash off unbound DNA/protein/salt/nucleotides and elute the RNA.

What type of sample can be used with TaqMan MicroRNA Assays?

One to ten nanograms of total RNA per 15 µL RT reaction is recommended for use with the TaqMan MicroRNA Assay. We have successfully detected miRNAs in as little as 25 pg of total RNA (~100 cells) in samples where the miRNA was highly abundant. The total RNA isolation procedure used must preserve small RNA species. We recommend the Invitrogen mirVana miRNA Isolation Kit (Cat. No. AM1560) or TRI reagent (Cat. No. AM9738).

When using the mirVana miRNA Isolation Kit, with phenol (Cat. No. AM1560), can I store homogenized tissue RNA lysate in the lysis buffer at -80 degrees C?

Yes, the RNA lysates can be stored at -80 degrees C for up to 2 months.

What kit do you recommend to capture small RNA that is &gt;17 nt in size?

We recommend using the mirVana miRNA Isolation Kit (Cat. No. AM1560, AM1561), both of which are designed to capture small RNAs, such as miRNAs, tRNAs, etc. Purification of RNAs as small as 20 nt has been obtained in-house using these kits. Note: Cat. No. AM1561 does not contain phenol, but is required for the procedure. Phenol can be purchased separately (Cat. No. AM9720).

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2792006Certificate of AnalysisJan 29, 2025AM1560
2791963Certificate of AnalysisJan 29, 2025AM1560
2800917Certificate of AnalysisJan 29, 2025AM1560
2804981Certificate of AnalysisJan 29, 2025AM1560
2792044Certificate of AnalysisJan 29, 2025AM1560
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (6)

Citations & References
Abstract
[Differential expression profile of microRNA between hyperplastic scar and normal skin].
Authors:Ning P, Liu DW, Mao YG, Peng Y, Lin ZW, Liu DM,
Journal:Zhonghua Yi Xue Za Zhi
PubMed ID:22781298
To explore the miRNA differential expression profiles of hyperplastic scar and normal skin so as to further elucidate the pathogenesis of hyperplastic scar and search for new therapeutic targets. The total RNA was extracted from 5 human hyperplastic scar and normal skin tissues by Trizol. The specimens were collected from ... More
Circulating microRNAs in serum: novel biomarkers for patients with bladder cancer?
Authors:Scheffer AR, Holdenrieder S, Kristiansen G, von Ruecker A, Müller SC, Ellinger J,
Journal:World J Urol
PubMed ID:23266581
'PURPOSE: Recent studies indicate that circulating microRNAs in serum/plasma are a novel class of non-invasive biomarkers with diagnostic and prognostic information. So far, circulating microRNAs have not been analyzed in patients with bladder cancer. METHODS: We collected serum from patients with non-muscle invasive bladder cancer (NMIBC), muscle invasive bladder cancer ... More
[Isolation, identification and analysis of the expression profile of miRNAs in Aedes albopictus].
Authors:Zheng PM, Wu JY, Gu JB, Tu ZJ, Chen XG,
Journal:Nan Fang Yi Ke Da Xue Xue Bao
PubMed ID:20423824
'To verify the miRNA in Aedes albopictus and characterize the expression profile of several miRNAs across all the life stages of Aedes albopictus. Based on the published miRNA sequences of Anopheles gambiae and Aedes aegypti, 6 DIG-labeled antisense probes were synthesized. The total RNAs from Aedes albopictus in 6 developmental ... More
MicroRNA expression profiles in the progression of prostate cancer-from high-grade prostate intraepithelial neoplasia to metastasis.
Authors:Leite KR, Tomiyama A, Reis ST, Sousa-Canavez JM, Sañudo A, Camara-Lopes LH, Srougi M,
Journal:Urol Oncol
PubMed ID:21880514
'Models of the multistep process related to cancer progression have been designed for many cancers including prostate. The aim of this study is to propose a new model including a possible role for recently described micro RNAs in prostate cancer (CaP) progression. Sixty-three patients underwent radical prostatectomy to treat localized ... More
High resolution of microRNA signatures in human whole saliva.
Authors:Patel RS, Jakymiw A, Yao B, Pauley BA, Carcamo WC, Katz J, Cheng JQ, Chan EK,
Journal:Arch Oral Biol
PubMed ID:21704302
Identifying discriminatory human salivary RNA biomarkers reflective of disease in a low-cost non-invasive screening assay is crucial to salivary diagnostics. Recent studies have reported both mRNA and microRNA (miRNA) in saliva, but little information has been documented on the quality and yield of RNA collected. Therefore, the aim of the ... More
6 total citations

Other products to consider



Share catalog number, name or link