Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

ULTRAhyb™ Ultrasensitive Hybridization Buffer

Catalog number: AM8670
Have Questions?
Invitrogen™

ULTRAhyb™ Ultrasensitive Hybridization Buffer

Catalog number: AM8670
Have Questions?
Catalog Number
AM8670
Unit Size
125 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 125 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM8670125 mL
-
AM86694 x 125 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Ambion™ ULTRAhyb™ contains a unique blend of hybridization accelerators and blocking agents that greatly enhance the levels of hybridization so that signals that once took days to visualize are now apparent in hours. It is supplied in one bottle containing 125 mL.

• Increase the sensitivity of any blot hybridization experiment
• Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
• Use less probe per blot
• Decrease hybridization time to just 2 hours

Increase Sensitivity Up to 100x or Decrease Hybridization to Just 2 Hours
Using standard hybridization buffers, only 1–5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19). With ULTRAhyb™ Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected.

For Any Hybridization Protocol
ULTRAhyb™ is compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots. It contains 50% formamide and is compatible with positively charged membranes (see Accessory Products). ULTRAhyb™ requires 10–100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction.

Accessory Products:
The BrightStar™ -Plus Positively Charged Nylon Membranes (SKU# AM10100, AM10102, or AM10104) are recommended for use with ULTRAhyb™. NorthernMax™ Wash Buffers (SKU# AM8673 and AM8674) are ideal for use with ULTRAhyb™.ULTRAhyb™-Oligo (SKU# AM8663) is specifically formulated for oligonucleotide probes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Hybridization Buffer
Product Type
Ultrasensitive Hybridization Buffer
Quantity
125 mL
Shipping Condition
Room Temperature

Contents & Storage

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text