Invitrogen™

DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Catalog number: D22065
Have Questions?
Invitrogen™

DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Catalog number: D22065
Have Questions?
Catalog Number
D22065
Unit Size
10 x 10 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D2206510 x 10 mg
-
D65675 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
DiFMUP is a fluorinated 4-methylumbelliferyl phosphate (MUP) derivative that has a lower pKa than that of MUP, making DiFMUP an excellent substrate for continuously assaying acid phosphatases at low pH. The reaction product of DiFMUP has excitation/emission maxima of ∼358/450 nm. DiFMUP is also available packaged in ten vials of 10 mg each (D-22065).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Blue
Detection Method
Fluorescence
Form
Powder
Quantity
10 x 10mg
Substrate
Phosphatase Substrate
Substrate Properties
Chemical Substrate
Target Enzyme
Phosphatases
Excitation/Emission
358⁄455
For Use With (Equipment)
Microplate Reader
Label or Dye
DiFMU
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text