Invitrogen™

DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Catalog number: D6567
Have Questions?
Invitrogen™

DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Catalog number: D6567
Have Questions?
Catalog Number
D6567
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 10 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D65675 mg
-
D2206510 x 10 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
DiFMUP is a fluorinated 4-methylumbelliferyl phosphate (MUP) derivative that has a lower pKa than that of MUP, making DiFMUP an excellent substrate for continuously assaying acid phosphatases at low pH. The reaction product of DiFMUP has excitation/emission maxima of ∼358/450 nm. DiFMUP is also available packaged in ten vials of 10 mg each (D-22065).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Blue
Detection Method
Fluorescent
Form
Powder
Quantity
5mg
Substrate
Phosphatase Substrate
Substrate Properties
Chemical Substrate
Target Enzyme
Phosphatases
Excitation/Emission
358⁄455
For Use With (Equipment)
Microplate Reader
Label or Dye
DiFMU
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text