BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine)
BODIPY&trade; FL C<sub>5</sub>-Sphingomyelin (<i>N</i>-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine)
Invitrogen™

BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine)

BODIPY™ FL C5スフィンゴミエリンはスフィンゴ脂質の輸送メカニズムおよび代謝メカニズムで使用できます詳細を見る
製品番号(カタログ番号)数量
D3522250 μg
製品番号(カタログ番号) D3522
価格(JPY)
81,700
Each
お問い合わせください ›
数量:
250 μg
Ask our AI about this Product
BODIPY™ FL C5スフィンゴミエリンはスフィンゴ脂質の輸送メカニズムおよび代謝メカニズムで使用できます。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
化学物質名または材質Sphingolipids
推奨保存方法Store in freezer (-5 to -30°C) and protect from light.
物理的フォームSolid
製品ラインBODIPY
数量250 μg
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

蛍光スペクトル

Fluorescence spectra

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
3061372Certificate of Analysis2024年10月09日D3522
2971120Certificate of Analysis2024年6月04日D3522
2785495Certificate of Analysis2023年8月30日D3522
2448078Certificate of Analysis2022年1月26日D3522
2281175Certificate of Analysis2020年12月21日D3522
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

引用および参考文献 (39)

引用および参考文献
Abstract
Fluorescence-based selection of gene-corrected hematopoietic stem and progenitor cells from acid sphingomyelinase-deficient mice: implications for Niemann-Pick disease gene therapy and the development of improved stem cell gene transfer procedures.
Authors:Erlich S, Miranda SR, Visser JW, Dagan A, Gatt S, Schuchman EH
Journal:Blood
PubMed ID:9864149
'The general utility of a novel, fluorescence-based procedure for assessing gene transfer and expression has been demonstrated using hematopoietic stem and progenitor cells. Lineage-depleted hematopoietic cells were isolated from the bone marrow or fetal livers of acid sphingomyelinase-deficient mice, and retrovirally transduced with amphotropic or ecotropic vectors encoding a normal ... More
Changes in the spectral properties of a plasma membrane lipid analog during the first seconds of endocytosis in living cells.
Authors:Chen CS, Martin OC, Pagano RE
Journal:Biophys J
PubMed ID:8994591
'N-[5-(5, 7-dimethyl Bodipy)-1-pentanoyl]-D-erythro-sphingosylphosphorylcholine (C5-DMB-SM), a fluorescent analog of sphingomyelin, has been used in a study of the formation of very early endosomes in human skin fibroblasts. This lipid exhibits a shift in its fluorescence emission maximum from green (approximately 515 nm) to red (approximately 620 nm) wavelengths with increasing concentrations ... More
Structural and functional regulation of tight junctions by RhoA and Rac1 small GTPases.
Authors:Jou TS, Schneeberger EE, Nelson WJ
Journal:J Cell Biol
PubMed ID:9660866
'Tight junctions (TJ) govern ion and solute diffusion through the paracellular space (gate function), and restrict mixing of membrane proteins and lipids between membrane domains (fence function) of polarized epithelial cells. We examined roles of the RhoA and Rac1 GTPases in regulating TJ structure and function in MDCK cells using ... More
Measurement of spontaneous transfer and transbilayer movement of BODIPY-labeled lipids in lipid vesicles.
Authors:Bai J, Pagano RE
Journal:Biochemistry
PubMed ID:9220970
'An assay was developed to study the spontaneous transfer and transbilayer movement (flip-flop) of lipid analogs labeled with the fluorescent fatty acid, 5-(5,7-dimethyl BODIPY)-1-pentanoic acid (C5-DMB-) in large unilamellar lipid vesicles comprised of 1-palmitoyl-2-oleoyl phosphatidylcholine (POPC). The assay is based on the concentration-dependent changes in fluorescence intensity that occur when ... More
The recycling endosome of Madin-Darby canine kidney cells is a mildly acidic compartment rich in raft components.
Authors:Gagescu R, Demaurex N, Parton RG, Hunziker W, Huber LA, Gruenberg J
Journal:Mol Biol Cell
PubMed ID:10930469
'We present a biochemical and morphological characterization of recycling endosomes containing the transferrin receptor in the epithelial Madin-Darby canine kidney cell line. We find that recycling endosomes are enriched in molecules known to regulate transferrin recycling but lack proteins involved in early endosome membrane dynamics, indicating that recycling endosomes are ... More
39 total citations

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection