Invitrogen™

BODIPY™ TR Cadaverine (5-(((4-(4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-yl)phenoxy)acetyl)amino)pentylamine, Hydrochloride)

Catalog number: D6251
Have Questions?
Invitrogen™

BODIPY™ TR Cadaverine (5-(((4-(4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-yl)phenoxy)acetyl)amino)pentylamine, Hydrochloride)

Catalog number: D6251
Have Questions?
Catalog Number
D6251
also known as D-6251
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D6251
also known as D-6251
5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The amine-containing BODIPY™ TR cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). The electronically neutral BODIPY™ TR dye is spectrally similar to the red fluorescent Texas Red™™ dye.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
5 mg
Product Type
TR Cadaverine
Chemical Reactivity
Carboxylic Acid, Ketone, Aldehyde
Shipping Condition
Room Temperature
Reactive Moiety
Amine, Cadaverine
Label or Dye
BODIPY™ TR
Label Type
BODIPY Dyes
Product Line
BODIPY™

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text