ELISA Assay Buffer (5X)
ELISA Assay Buffer (5X)
Invitrogen™

ELISA Assay Buffer (5X)

ELISA Assay Buffer (5X) is intended for use as: blocking reagent for ELISA plates, diluent for ELISA standards and samples,Read more
Have Questions?
Catalog NumberQuantity
DS98200200 mL
Catalog number DS98200
Price (USD)
219.65
Online Exclusive
229.00
Save 9.35 (4%)
Each
In stock
Add to cart
Quantity:
200 mL
Recurring order eligible. Learn more »
Price (USD)
219.65
Online Exclusive
229.00
Save 9.35 (4%)
Each
Add to cart
Ask our AI about this Product
ELISA Assay Buffer (5X) is intended for use as: blocking reagent for ELISA plates, diluent for ELISA standards and samples, diluent for the detection antibody, and diluent for the HRP conjugate. This buffer is recommended when using Invitrogen Antibody Pairs or when developing a sandwich ELISA.

For a complete kit of ELISA buffers, see ELISA Buffer Kit.
View all available buffers and reagents for ELISA applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping ConditionWet Ice
ApplicationsELISA
FormConcentrated
Quantity200 mL
Unit SizeEach
Contents & Storage
Buffer. Store at 2-8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How do I develop a sandwich ELISA using Cytosets?

Each CytoSets contains capture (coating) antibody, biotinlyated detection antibody, standard and Streptavidin-HRP. Other reagents required are listed in the CytoSets information sheet included with the kit and can be purshased from us separately (Antibody Pair Buffer kit CNB0011, 5x Assay Buffer DS98200, etc.). The information sheet also provides a specific procedure and illustrates an example standard curve which can be obtained when the specific procedure is followed. A general procedure is summarized here:

1) Coat the microplate with diluted capture (coating) antibody overnight at 2-8 degrees C; Wash the plate
2) Incubate standards or samples with the coated microplate; Wash the plate
3) Incubate diluted biotinlyated detection antibody with the plate; Wash the plate
4) Incubate Streptavidin-HRP with the plate for 15-45 minutes; Wash the plate
5) Incubate the plate with TMB substrate for 10-60 minutes and stop the reaction with Stop solution
6) Read microplate at 450 nm.

Investigators are advised to determine optimal buffer formulations, concentrations and incubation times for individual applications.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
456931-000Certificate of AnalysisJul 11, 2025DS98200
449766-000Certificate of AnalysisJul 03, 2025DS98200
449765-000Certificate of AnalysisJul 03, 2025DS98200
441945-000Certificate of AnalysisJun 14, 2025DS98200
445706-000Certificate of AnalysisJun 14, 2025DS98200
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (1)

Citations & References
Abstract
Neutrophil CD11b, L-selectin and Fc IgA receptors in patients with dermatitis herpetiformis.
Authors:Smith AD, Streilein RD, Hall RP,
Journal:Br J Dermatol
PubMed ID:12452859
'The skin lesions found in patients with dermatitis herpetiformis (DH) are characterized by the presence of neutrophils at the dermal papillary tips in areas where the diagnostic cutaneous IgA deposits are found. Although the presence of the skin lesions of DH is known to be associated with gluten-sensitive enteropathy, the ... More
1 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection