ELISA Stop Solution is a ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution. As TMB substrate solution reacts with horseradish peroxidase, it produces a pronounced blue color. At the end of TMB incubation, a volume of ELISA Stop Solution equal to the volume of TMB substrate solution is added to the wells to terminate the horseradish peroxidase reaction. Addition of Stop Solution results in the rapid formation of a yellow end product. The optical density of this yellow end product is measured at 450 nm. The yellow end product is stable for sixty minutes after addition of Stop Solution and so optical density should be measured within that time.
For Research Use Only. Not for use in diagnostic procedures.
規格
FormLiquid
Shipping ConditionAmbient
Substrate TypeHRP, TMB
Product TypeELISA Stop Solution
Quantity1 L
Unit Size1 L
內容物與存放
Store at 2°C to 8°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.