T4 DNA Ligase (5 U/μL)
T4 DNA Ligase (5 U/μL)
Thermo Scientific™

T4 DNA Ligase (5 U/μL)

Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplexRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EL00125 x 1,000 units
EL00111,000 units
EL0014200 units
Catalog number EL0012
Price (USD)
903.65
Online Exclusive
1,106.00
Save 202.35 (18%)
Each
-
Add to cart
Quantity:
5 x 1,000 units
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
903.65
Online Exclusive
1,106.00
Save 202.35 (18%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific T4 DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA. The enzyme repairs single-strand nicks in duplex DNA, RNA, or DNA/RNA hybrids. It also joins DNA fragments with either cohesive or blunt termini, but has no activity on single-stranded nucleic acids.

T4 DNA Ligase requires ATP as a cofactor.

Highlights

• Active in Themo Scientific restriction enzyme, PCR, and RT buffers (when supplemented with ATP)
• Fast—sticky-end ligation is completed in 10 minutes at room temperature
• Supplied with PEG solution for efficient blunt-end ligation

Applications

• Cloning of restriction enzyme generated DNA fragments
• Cloning of PCR products
• Joining of double-stranded oligonucleotide linkers or adaptors to DNA
• Site-directed mutagenesis
• Amplified fragment length polymorphism (AFLP)
• Ligase-mediated RNA detection (see Reference 3)
• Nick repair in duplex DNA, RNA or DNA/RNA hybrids
• Self-circularization of linear DNA.

Includes

• T4 DNA Ligase
• 10X T4 DNA Ligase Buffer
• 50% PEG Solution

Notes

• Binding of T4 DNA Ligase to DNA may result in a band shift in agarose gels. To avoid this, incubate samples with 6X DNA Loading Dye & SDS Solution at 70°C for 5 min or 65°C for 10 minutes and chill on ice prior to electrophoresis.
• The volume of the ligation reaction mixture should not exceed 10% of the competent cell volume in the transformation process.
• Prior to electro-transformation, remove T4 DNA Ligase from the ligation mixture using spin columns or chloroform extraction. The extracted DNA can be further precipitated with ethanol.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X T4 DNA Ligase Buffer
Product TypeT4 DNA Ligase
Quantity5 x 1,000 units
Concentration5 U/μL
EnzymeDNA Ligase
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3264898Certificate of AnalysisJun 19, 2025EL0012
3262948Certificate of AnalysisJun 18, 2025EL0014
3262939Certificate of AnalysisJun 17, 2025EL0014
3262855Certificate of AnalysisJun 17, 2025EL0012
3262534Certificate of AnalysisJun 17, 2025EL0011
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The white precipitates are DTT in the buffer which may appear after freezing. To redissolve DTT, warm up the buffer to 37 degrees C by holding the vial between your palms, or incubating in a water bath for a few minutes, and mix well. The formation of DTT precipitates does not influence the quality of the buffer. The 10X T4 DNA Ligase Buffer is also available separately (Cat. No. B69).

The speed of ligation is the main difference between the two. The Thermo Scientific Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minutes at room temperature. The kit contains T4 DNA ligase and a specially-formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation.

Thermo Scientific T4 DNA ligase activity is expressed in Weiss units. One Weiss unit is equivalent to approximately 200 cohesive end units (CEU) which may be used by other vendors. One Weiss unit of the enzyme catalyzes the conversion of 1 nmol of [32PPi] into Norit-adsorbable form in 20 min at 37 degrees C. One CEU is defined as the amount of enzyme required to give 50% ligation of HindIII fragments of lambda DNA in 30 min at 16 degrees C.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection