Thermo Scientific™
Electrical Feed-through Sample Holder
Catalog number: ELFEEDTHRUSAMPLHLD
Have Questions?
Thermo Scientific™
Electrical Feed-through Sample Holder
Catalog number: ELFEEDTHRUSAMPLHLD
Have Questions?
The Thermo Scientific™ Electrical Feed-through Sample Holder allows users to connect electrical probes to the sample for in-situ measurements. Six quick release pin connections can be used for various signals to be read and voltages or currents to be applied to the sample simultaneously while the sample is observed in the SEM.
Catalog Number
ELFEEDTHRUSAMPLHLD
Unit Size
Each
Price (USD)
Full specifications
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
ELFEEDTHRUSAMPLHLDFull specifications
EachRequest A Quote
Unit SizeEach
Showing 1 of 1
The sample can be height adjusted from 0–25 mm manually to accommodate a wide variety of samples on a 12 mm pin stub.
Applications
  • Activate Mems devices
  • Measuring probe current