Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Endonuclease V, T.maritima (5 U/μL)

Catalog number: EN0141
Have Questions?
Thermo Scientific™

Endonuclease V, T.maritima (5 U/μL)

Catalog number: EN0141
Have Questions?
Catalog Number
EN0141
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EN0141250 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Endonuclease V, T. maritima (Endo V) is a 3'-endonuclease involved in DNA repair, which initiates removal of deaminated bases from damaged DNA, including uracil, hypoxanthine, and xanthine.

Endonuclease V is also active toward abasic sites and urea sites, base pair mismatches, flap and pseudo Y structures, and small insertions/deletions in DNA molecules. The cleavage site generated by Endonuclease V is at the second phosphodiester bond 3' to a lesion.

Highlights

• Optimal activity at temperatures of 65 to 70°C

Applications

• High-throughput methods for mutation research
• Studies in mutagenesis and DNA repair
• Mismatch cleavage
• Genotyping

Note

Use of this enzyme in certain applications may be covered by patents and may require a license.

When the enzyme is in excess, the primary nicked products experience a second nicking event on the complementary strand, leading to a double-stranded break. At low concentrations, however, Endonuclease V first nicks a DNA strand at the lesions located closer to the 5'-end of DNA molecule. Single-stranded DNA is cleaved with much lower efficiency. Mg2+ or Mn2+ ions are required for enzyme activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5 U/μL
Enzyme
Endonuclease
Quantity
250 U
Product Type
Endonuclease V

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text