Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Klenow Fragment, LC (2 U/μL)

Catalog number: EP0054
Have Questions?
Thermo Scientific™

Klenow Fragment, LC (2 U/μL)

Catalog number: EP0054
Have Questions?
Catalog Number
EP0054
Unit Size
300 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EP0054300 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.

Highlights

• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
• Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers

Applications
• DNA blunting by fill-in 5'-overhangs
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA
• DNA sequencing by the Sanger method
• Site-specific mutagenesis of DNA with synthetic oligonucleotides
• Second strand synthesis of cDNA
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
2 U/μL
Polymerase
DNA Polymerase I
Quantity
300 U
Product Type
Klenow Fragment

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text