Thermo Scientific™
Eucentric Sample Holder
Catalog number: EUCSAMPLEHOLDER
In many SEM applications, a user can gain more insight into sample properties if the sample can be tilted and rotated. The Thermo Scientific™ Eucentric Sample Holder for the Phenom XL has been specifically developed with that in mind. The holder contains a sub-stage that allows users to easily and safely look at a sample from all sides.