Invitrogen™

Anza™ T4 DNA Ligase Master Mix

Catalog number: IVGN2108
Have Questions?
Invitrogen™

Anza™ T4 DNA Ligase Master Mix

Catalog number: IVGN2108
Have Questions?
Catalog Number
IVGN2108
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
IVGN2108200 reactions
-
IVGN210450 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen Anza™ T4 DNA Ligase Master Mix facilitates the joining of abutted 5’-phosphate and 3’-hydroxyl termini in duplex DNA through the formation of a phosphodiester bond. It can be used to join DNA fragments with both sticky ends and blunt ends, and to repair nicks in double-stranded DNA with 3'-hydroxyl and 5'-phosphate ends. Anza T4 DNA Ligase is formulated as a 4X concentrated master mix. Ligation can be performed with DNA in water, TE, elution buffer, or 1X Anza™ buffers.

Benefits:
• Ligation complete in 15 minutes at room temperature
• A single ligase master mix for both sticky-end and blunt-end ligation reactions
• Ready-to-use master mix format reduces pipetting steps
• 4X concentration enables more dilute DNA to be used in ligation reaction without concentrating

Anza T4 DNA Ligase Master Mix is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
4X
Shipping Condition
Approved for shipment on Wet or Dry Ice
Enzyme
T4 DNA Ligase
Compatible Buffer
Elution Buffer, 1X Anza Buffer
Quantity
200 Reactions
Product Type
T4 DNA Ligase Master Mix
Product Line
Anza™

Contents & Storage

1 mL Anza T4 DNA Ligase Master Mix

Store at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text