Invitrogen™

Anza™ Alkaline Phosphatase

Catalog number: IVGN2208
Have Questions?
Invitrogen™

Anza™ Alkaline Phosphatase

Catalog number: IVGN2208
Have Questions?
Catalog Number
IVGN2208
Unit Size
2000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
IVGN22082000 reactions
-
IVGN2204500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Invitrogen™ Anza™ Alkaline Phosphatase is intended for use in the removal of 3' and 5' phosphate groups that remain after Anza™ restriction enzyme digestion, such as for the dephosphorylation of vectors prior to insert ligation and removal of 5′ phosphates prior to end-labeling. Since the 5′ phosphate group is required for DNA ligation, treatment of vectors with Anza Alkaline Phosphatase prevents self-ligation and re-circularization, resulting in decreased vector background when cloning.

Features of Anza Alkaline Phosphatase:

• Validated for use with all Anza restriction enzymes
• Dephosphorylates in 15 minutes at 37°C
• Heat inactivated after 5 minutes at 80°C
• Fully active in Anza Buffer
• Dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs)

Flexible protocol
Some restriction enzymes prohibit or reduce the dephosphorylation process when digestion and dephosphorylation are performed simultaneously. Therefore, Anza Alkaline Phosphatase may be used in one of three ways depending upon the restriction enzyme used:

One-Step Protocol: Simultaneous restriction enzyme digestion and dephosphorylation in Anza buffer
Heat Inactivation Two-Step Protocol: Digestion is carried out first, followed by heat inactivation (20 minutes at 80oC) then dephosphorylation
Column Purification Two-Step Protocol: Digestion is carried out first, followed by column purification then dephosphorylation

No need for enzyme removal
Anza Alkaline Phosphatase is completely inactivated after 5 minutes at 80°C, so enzyme removal via phenol/chloroform extraction or column purification is not required prior to subsequent ligation steps. If the One-Step protocol was used, heat the reaction mixture for 20 minutes at 80oC to inactivate the restriction enzyme as well as the alkaline phosphatase. Heat inactivation of the alkaline phosphatase prevents residual phosphatase from dephosphorylating the DNA fragments added to the reaction mixture for ligation.

Anza™ Alkaline Phosphatase is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza T4 PNK Kit, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit.

For more information on the Anza system, visit www.thermofisher.com/anza.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Approved for shipment on Wet or Dry Ice
Enzyme
Alkaline Phosphatase
Compatible Buffer
10X Buffer
Quantity
2000 Reactions
Product Type
Alkaline Phosphatase
Product Line
Anza™

Contents & Storage

2 mL Anza Alkaline Phosphatase
1 mL Anza Buffer (10X)
1 mL Anza Red Buffer (10X)

Store at -5 to -30°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text