Thermo Scientific Chemicals

Peroxidase, horseradish

Catalog number: J60026.MC
100 mg, Each
Thermo Scientific Chemicals

Peroxidase, horseradish

Catalog number: J60026.MC
100 mg, Each
Quantity
Catalog number: J60026.MC
also known as J60026-MC
Price (USD)
Quantity
-
Have Questions?
Assay (special)
RZ (Reinheitszahl): ≥1.0% (absorbance ratio A403/A275 determined at 1.0 mg/ml in deionized water)
Form
Lyophilized powder
units/mgdw
≥85 units per mg dry weight
Appearance (Color)
Brown to dark brown and red to dark red
Unit definition
One unit decomposes one micromole of hydrogen peroxide per minute at 25°C, pH 7.0, using a 4-aminoantipyrine/phenol assay at 510nm

Description

Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text