Thermo Scientific Chemicals

Ribonuclease T1, Aspergillus oryzae in 2.8M ammonium sulfate

Catalog number: J61644.EXP
100 kU, Each
Thermo Scientific Chemicals

Ribonuclease T1, Aspergillus oryzae in 2.8M ammonium sulfate

Catalog number: J61644.EXP
100 kU, Each
Quantity
Catalog number: J61644.EXP
also known as J61644-!P
Price (USD)
Quantity
-
Have Questions?
Unit definition
One unit causes an increase in absorbance of 1.0 at 260 nm at 37?C, pH 7.5 in 15 minutes.
Form
Supplied as solution in 2.8 M ammonium sulfate
units/mgP
≥300,000 units/ mg protein
Concentration v/v
Units / ml: ≥ 100,000

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text