TE buffer, (20X), pH 8.0, autoclaved
TE buffer, (20X), pH 8.0, autoclaved
TE buffer, (20X), pH 8.0, autoclaved
TE buffer, (20X), pH 8.0, autoclaved
Thermo Scientific Chemicals

TE buffer, (20X), pH 8.0, autoclaved

Commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids. | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number J62388.AP
also known as J62388-AP
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Add to cart
Specifications
CAS1185-53-1
Chemical Name or MaterialTE buffer
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
MDL NumberMFCD00236359
View more
Ask our AI about this Product
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA.  TE buffer functions to solubilize DNA or RNA while protecting it from degradation. This buffer is used at different pH values and has become the standard buffer for the storage of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
S22L521Certificate of AnalysisJun 10, 2025J62388.AP, J62388.AK
Q19L542Certificate of AnalysisMay 17, 2025J62388.AP, J62388.AK
Z28K538Certificate of AnalysisJan 21, 2025J62388.AP, J62388.AK
X20K551Certificate of AnalysisOct 30, 2024J62388.AP, J62388.AK
R18K507Certificate of AnalysisJun 14, 2024J62388.AP, J62388.AK
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection