TBE Buffer (Tris-borate-EDTA) (10X)
TBE Buffer (Tris-borate-EDTA) (10X)
Thermo Scientific™

TBE Buffer (Tris-borate-EDTA) (10X)

Thermo Scientific 10X TBE Buffer (Tris-borate-EDTA) is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBERead more
Have Questions?
Catalog NumberQuantity
B521 L
Catalog number B52
Price (USD)
63.65
Online Exclusive
73.25
Save 9.60 (13%)
Each
-
Add to cart
Quantity:
1 L
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
73.25
Save 9.60 (13%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific 10X TBE Buffer (Tris-borate-EDTA) is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels, but is not recommended for preparative gels for recovery of nucleic acids. Since borate in TBE buffer is a strong inhibitor for many enzymes, TAE buffer (Tris Acetate-EDTA buffer, 10X powder, sc-296647) is recommended when looking at enzymatic applications for the DNA sample.

Applications

• Electrophoresis of nucleic acids in agarose and polyacrylamide gels
• Used both as a running buffer and as a gel preparation buffer
• Filtered through a 0.22 μm membrane
• Recommended for electrophoresis of RNA and DNA fragments smaller than 1500 bp

Note
Double-stranded linear nucleic acid molecules migrate about 10% slower in TBE buffer than in TAE buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTBE Buffer
Quantity1 L
Concentration10X
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3256689Certificate of AnalysisJun 09, 2025B52
3203592Certificate of AnalysisApr 04, 2025B52
3203488Certificate of AnalysisApr 03, 2025B52
3202586Certificate of AnalysisApr 02, 2025B52
3193960Certificate of AnalysisMar 20, 2025B52
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

A TAE running buffer used in conjunction with a TBE gel would create high current and may lead to gel hydrolysis. Also, the acetate ion in TAE migrates faster than the borate ion in TBE buffer, and can create fuzzy bands. It is possible to try soaking the TBE gel (run in TBE buffer) in the TAE buffer following the run and then eluting from that. Since nucleic acids diffuse out of the gel relatively slowly, this may work.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection