Tylosin tartrate, 94%
Tylosin tartrate, 94%
Tylosin tartrate, 94%
Tylosin tartrate, 94%
Thermo Scientific Chemicals

Tylosin tartrate, 94%

Macrolide antibiotic | CAS: 1405-54-5 | C50H83NO23 | 1066.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62633.09
also known as J62633-09
Price (USD)
322.65
Special offer
Online exclusive
Ends: 26-Sep-2025
398.00
Save 75.35 (19%)
Each
In stock
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
322.65
Special offer
Online exclusive
Ends: 26-Sep-2025
398.00
Save 75.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS1405-54-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
Assay (unspecified)≥92.0%
Assay (unspecified)Tyramine: ≤0.35%
Sulfated Ash≤2.5%
Loss on Drying≤4.5%
View more
Ask our AI about this Product
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an antibacterial agent. This compound has been shown to be effective in vitro against Staphylococcus intermedius and sensitive mycoplasma organisms. Although a lot of antibiotics tend to suppress the immune system, macrolide antibiotics have been reported to enhance the immune system when used in chicken models. Studies suggest that chronic exposure to this compound may have a negative effect on the respiratory system. For turkeys - for treatment of infectious synovitis and sinusitis. It is effective also for treatment of ileitis, dysentery and necrotic enteritis in swine. It is successfully applied as a preventive measure in stress conditions as vaccinations, transportation and others.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Knothe H. A review of the medical considerations of the use of tylosin and other macrolide antibiotics as additives in animal feeds.Infection.,1977,5(3), 183-7.
  2. Lee HS, et al. Occupational asthma due to tylosin tartrate.Br J Ind Med.,1989,46(7), 498-9.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
P19L039Certificate of AnalysisMar 21, 2025J62633.03, J62633.09, J62633.06
N03L024Certificate of AnalysisFeb 05, 2025J62633.03, J62633.09, J62633.06
U12K008Certificate of AnalysisAug 28, 2024J62633.03, J62633.09, J62633.06
R14K042Certificate of AnalysisMay 21, 2024J62633.03, J62633.09, J62633.06
M16G031Certificate of AnalysisMar 14, 2024J62633.03, J62633.09, J62633.06
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection