Thermo Scientific Chemicals

Hexokinase, yeast

Catalog number: J63795.JPL
10 kU, Each
Thermo Scientific Chemicals

Hexokinase, yeast

Catalog number: J63795.JPL
10 kU, Each
Quantity
Catalog number: J63795.JPL
also known as J63795-J+
Price (USD)
Quantity
-
Have Questions?
Form
Lyophilized powder
Assay (special)
Contaminating activities: Phosphohexos isomerase: ≤ 0.10%; 6-phosphogluconate dehydrogenase: ≤0.005%; Adenylate kinase: ≤0.005%; Creatine phosphokinase: ≤0.005%
units/mgP
≥ 150
Unit definition
One unit reduces one micromole of NAD per minute at 30?C, pH 8.0
units/mgdw
Report value

Description

Hexokinase is used to catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing, ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text