Maxima H Minus First Strand cDNA Synthesis Kit
Maxima H Minus cDNA Synthesis Master Mix now available. Get a sample »
Maxima H Minus First Strand cDNA Synthesis Kit
Thermo Scientific™

Maxima H Minus First Strand cDNA Synthesis Kit

Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesNo. of Reactions
K1682Kit with dsDNase100 Reactions
K1651Kit only20 Reactions
K1652Kit only100 Reactions
K1681Kit with dsDNase20 Reactions
Catalog number K1682
Price (USD)
690.65
Online Exclusive
834.00
Save 143.35 (17%)
Each
-
Add to cart
Includes:
Kit with dsDNase
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
690.65
Online Exclusive
834.00
Save 143.35 (17%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Maxima H Minus First Strand cDNA Synthesis Kit, available with or without dsDNase, is a complete system for highly efficient synthesis of first strand cDNA. The kit uses the Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT. The enzyme features the highest thermostability among the derivatives of MMLV RT and lacks RNase H activity.

This kit allows synthesis of long cDNAs up to 20 kb at elevated temperatures (up to 65°C), superseding other systems' abilities to produce full-length cDNA. Due to increased synthesis rates the reaction can be completed in 30 minutes.

For reverse transcription the kit uses Maxima H Minus Reverse Transcriptase (RT), which is an advanced enzyme derived by in vitro evolution of MMLV RT.

The Maxima H Minus First Strand cDNA Synthesis Kit with dsDNase provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. The kit contains a novel double-strand-specific DNase (dsDNase) engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific dsDNase activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved, and dsDNase treated RNA can be directly added to reverse transcription.

Features of the Maxima H Minus First Strand cDNA Synthesis Kit include:
• Increased reaction temperatures—the first strand of cDNA can be synthesized within the 42–65°C temperature range
• High yields of full-length first strand cDNA—with RNA templates up to 20 kb
• Flexible priming—oligo(dT)18, random hexamer or gene-specific primers
• Integrated genomic DNA removal step with dsDNase kit

Applications
• First Strand cDNA synthesis for RT-PCR
• Construction of cDNA libraries
• Generation of probes for hybridization
• Antisense RNA synthesis

Contents
• Maxima H Minus Enzyme Mix
• Oligo(dT)18 and Random hexamer primers
• 5X RT Buffer
• dNTP Mix
• Nuclease-free water

First Strand Synthesis Kit with dsDNase also contains
• dsDNase and 10X dsDNase Buffer

Additional information about reaction components
• Maxima H Minus Enzyme Mix contains Maxima H Minus Reverse Transcriptase and RiboLock RNase Inhibitor. RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
• Oligo(dT)18 and random hexamer primers are supplied with the kit. Random hexamer primers bind non-specifically and are used to synthesize cDNA from all RNAs in a total RNA population. The oligo(dT)18 primer selectively anneals to the 3'-end of poly(A) RNA, synthesizing cDNA only from poly(A) tailed mRNA. Gene-specific primers may also be used with the kit to prime synthesis from a specified sequence.
• 10 mM dNTP Mix is a premixed aqueous solution of dATP, dTTP, dCTP, and dGTP.
• Nuclease-free water is provided for reaction set-up and dilution of sample DNA. The absence of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatKit
IncludesKit with dsDNase
No. of Reactions100 Reactions
Optimal Reaction Temperature50°C to 55°C
QuantityEach
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseMaxima H Minus
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
Size (Final Product)Up to 20 kb
Starting MaterialRNA
TechniqueReverse Transcription
For Use With (Application)RT-PCR
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• Maxima H Minus Enzyme Mix
• dsDNase
• 10X dsDNase Buffer
• Oligo(dT)18 and Random hexamer primers
• 5X RT Buffer
• dNTP Mix
• Nuclease-free water

Store at –20°C.

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3274410Certificate of AnalysisJul 04, 2025K1651
3274434Certificate of AnalysisJul 04, 2025K1651
3271432Certificate of AnalysisJul 01, 2025K1651
3271461Certificate of AnalysisJul 01, 2025K1651
3271144Certificate of AnalysisJun 30, 2025K1682
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

dsDNase can be inactivated by incubating the sample at 55°C for 5 min in the presence of 10 mM DTT.

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. It is also recommended to use RNase H-minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in qPCR applications.

All Thermo Scientific reverse transcriptases possess intrinsic TdT activity although at varying degrees depending upon the reaction conditions. For addition of template-independent C nucleotides (as for SMART and RACE experiments), this specific TdT activity can be induced by Mn2+. We would recommend Maxima H- or RevertAid H- minus RTs for this purpose.

No, dsDNase cleaves only double-stranded DNA. However, if ssDNA forms double-stranded structures, it will act as a target for dsDNase. Because of this, long and complex ssDNA may be partially degraded. Therefore we recommend additional dsDNase inactivation step for RT-PCR amplification of targets 3 kb or longer. The inactivation step should be performed by 5 min incubation at 55 degrees C in the presence of 10 mM DTT.

No. RT reaction composition inhibits dsDNase activity, and genomic DNA removal may be incomplete.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection