Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™
RapidOut DNA Removal Kit
Catalog number: K2981
Have Questions?
Thermo Scientific™
RapidOut DNA Removal Kit
Catalog number: K2981
Have Questions?
Catalog Number
K2981
Unit Size
50 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K298150 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific RapidOut DNA Removal Kit rapidly and safely removes genomic DNA from total RNA and mRNA preparations. Complete digestion of DNA and safe removal of DNase I from the digestion reaction is ensured without RNA damaging steps, such as heating or organic extraction. First, the RNA sample is treated with recombinant RNase-free DNase I to levels below the limit of detection by routine PCR. DNase I is safely removed subsequently using proprietary DNase Removal Reagent (DRR).
DRR efficiently binds DNase I and the complex is collected at the bottom of the tube by centrifugation. The purified RNA is collected as a supernatant. The RNA after RapidOut procedure is free from DNA contamination and free of DNase I. It is ready to use in different applications including end-point or real-time RT-PCR, cloning, microarrays, and Northern blotting.
Highlights
Efficient—complete ds and ssDNA digestion and proprietary technology for DNase I removal
Rapid—single step sufficient for complete DNase I removal
Safe—no need for toxic organic extractions or RNA-damaging heating steps
Applications
Main Applications
• RNA isolation and RNA analysis, particularly RT-qPCR and RT-PCR (customers performing expression analysis of low transcription level genes.
• Customers performing ds-cDNA synthesis from total RNA preps.
Other applications
• Elimination of DNA from RNA for microinjections and transfection.
• Elimination of DNA from RNA prior microarray analysis.
• Elimination of DNA from RNA prior Northern blot analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Enzyme
DNase
Product Type
Removal Kit