Recirculating Chillers
Recirculating chillers allow for quick ramp-up and cool-down for applications such as separations, chemical reaction control, spectroscopy and laboratory automation. Products are available in a variety of achievable temperature ranges and reservoir volumes. Thermo Scientific™ Polar Series Accel 500 LT CoolingHeating Recirculating Chillers offer flexibility, accuracy, and efficiency offering a full-range of cooling capabilities for quick ramp-up and cool-down of your application.