Search Thermo Fisher Scientific
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
METRIOSTEM | - | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
High-volume TEM data, accurate and repeatable - at the lowest cost-per-sample
Metrios TEM automates the basic TEM operation and measurement procedures, minimizing the requirements for specialized operator training. Its advanced automated metrology routines deliver significantly greater precision than manual methods. The Metrios TEM is designed to provide customers with improved throughput and lower cost-per-sample than other TEMs.