Thermo Scientific™
Micro Tool Sample Holder
Catalog number: MICROTOOLSAMPLHOLD
Imaging long axial shaped samples without sample modification is possible with the Thermo Scientific™ Micro Tool Sample Holder. Imaging objects such as drill bits, milling tools and injection needles is fast and easy using the Micro Tool Sample Holder. Imaging coatings and cutting angles is possible by rotating and tilting the region of interest without unloading the sample from the holder.