Invitrogen™

Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 5-isomer

Catalog number: O6147
Have Questions?
Invitrogen™

Oregon Green™ 488 Carboxylic Acid, Succinimidyl Ester, 5-isomer

Catalog number: O6147
Have Questions?
Catalog Number
O6147
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
O61475 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This single 5-isomer preparation of our proprietary, amine-reactive dye, Oregon Green 488 carboxylic acid, succinimidyl ester and its conjugates have green fluorescence similar to that of fluorescein but are more photostable. Additionally, Oregon Green 488 dye has a lower pKa than fluorescein (pKa = 4.7 versus 6.4 for fluorescein), making this bright dye less pH sensitive in the physiological range.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
5 mg
Product Type
Carboxylic Acid Succinimidyl Ester
Chemical Reactivity
Amine
Shipping Condition
Room Temperature
Reactive Moiety
Active Ester, Succinimidyl Ester
Label or Dye
Oregon Green™ 488
Label Type
Classic Dyes
Product Line
Oregon Green™

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text