Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 Assays (2 mL assay volume) |
Packaging Type | 10 x 100 μL Tubes |
Quantitation Range | 50 pg to 2 μg |
Product Line | PICOGREEN, Quant-iT |
Product Type | Quant-iT PicoGreen dsDNA Assay Kit |
Shipping Condition | Room Temperature |
Unit Size | 10 x 100 µL kit |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 Assays (2 mL assay volume) |
Packaging Type | 10 x 100 μL Tubes |
Quantitation Range | 50 pg to 2 μg |
Product Line | PICOGREEN, Quant-iT |
Product Type | Quant-iT PicoGreen dsDNA Reagent |
Shipping Condition | Room Temperature |
Unit Size | 10 x 100 µL |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 Assays (2 mL assay volume) |
Packaging Type | 1 x 1 mL Bottle |
Quantitation Range | 50 pg to 2 μg |
Product Line | PICOGREEN, Quant-iT |
Product Type | Quant-iT PicoGreen dsDNA Reagent |
Shipping Condition | Room Temperature |
Unit Size | 1 mL |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 Assays (2 mL assay volume) |
Packaging Type | 1 x 1 mL Bottle |
Quantitation Range | 50 pg to 2 μg |
Product Line | PICOGREEN, Quant-iT |
Product Type | Quant-iT PicoGreen dsDNA Assay Kit |
Shipping Condition | Room Temperature |
Unit Size | 1 mL kit |
產品號碼 | 規格 | Unit Size | Product Type | Quantitation Range | Packaging Type | 價格 (HKD) | 供應狀況 | 數量 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
P11496 | 10 x 100 µL kit | Quant-iT PicoGreen dsDNA Assay Kit | 50 pg to 2 μg | 10 x 100 μL Tubes | 價格 5,574.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P11495 | 10 x 100 µL | Quant-iT PicoGreen dsDNA Reagent | 50 pg to 2 μg | 10 x 100 μL Tubes | 價格 5,214.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P7581 | 1 mL | Quant-iT PicoGreen dsDNA Reagent | 50 pg to 2 μg | 1 x 1 mL Bottle | 價格 4,783.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P7589 | 1 mL kit | Quant-iT PicoGreen dsDNA Assay Kit | 50 pg to 2 μg | 1 x 1 mL Bottle | 價格 5,432.00 您的價格 | *** | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|