Premo Autophagy Assays with Sensor LC3B, p62, or Tandem Sensor RFP-GFP-LC3B
Premo Autophagy Assays with Sensor LC3B, p62, or Tandem Sensor RFP-GFP-LC3B
Invitrogen™

Premo Autophagy Assays with Sensor LC3B, p62, or Tandem Sensor RFP-GFP-LC3B

Detect cell autophagy with our Autophagy Assay Kits, which offer chimeric sensors containing fluorescent proteins GFP and RFP, and autophagy-associated proteins p62 and LC3B.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorLabel or DyeDescription
P36240GreenGFP-p62Premo Autophagy Sensor GFP-p62 Kit
P36235GreenLC3B-GFPPremo Autophagy Sensor LC3B-GFP (BacMam 2.0)
P36236RedLC3B-RFPPremo Autophagy Sensor LC3B-RFP (BacMam 2.0)
P36239Red-Orange, GreenRFP-GFP-LC3BPremo Autophagy Tandem Sensor RFP-GFP-LC3B Kit
P36241RedRFP-p62Premo Autophagy Sensor RFP-p62 Kit
Catalog number P36240
Price (USD)
920.65
Online Exclusive
954.00
Save 33.35 (3%)
Each
In stock
Add to cart
Color:
Green
Label or Dye:
GFP-p62
Description:
Premo Autophagy Sensor GFP-p62 Kit
Recurring order eligible. Learn more »
Price (USD)
920.65
Online Exclusive
954.00
Save 33.35 (3%)
Each
Add to cart
Ask our AI about this Product
Easily monitor various stages of cellular autophagy with Premo Autophagy assays and sensors, which are based on LC3B- or p62-linked fluorescent protein chimeras, or the RFP-GFP-LC3B tandem sensor. Premo Autophagy assay kits offer the selectivity of Premo Autophagy sensors, which are based on chimeras of autophagy-associated proteins LC3B or p62, and green (GFP) and/or red fluorescent protein (RFP), with the transduction efficiency of BacMam technology. Transduction or even co-transduction can be performed in a wide variety of cells, including neurons and neural stem cells (NSCs) using a simple protocol that involves adding the BacMam 2.0 chimeric reagent to cells, incubating overnight, and then visualizing transduction using standard GFP (green fluorescent protein) and/or RFP (red fluorescent protein) settings.
Premo Autophagy sensors take advantage of autophagy-associated proteins LC3B or p62, along with GFP or RFP, to detect either the induction or inhibition of autophagy in cells. The Premo Autophagy sensors are transduced using BacMam (insect Baculovirus with a Mammalian promoter) technology, enabling unambiguous visualization of the chimeric proteins in autophagosomes. BacMam reagents are non-replicating in mammalian cells and therefore safe to handle. They are also non-cytotoxic and ready-to-use. Unlike typical expression vectors, BacMam reagents enable titratable and reproducible expression, and offer high co-transduction efficiency; therefore, multiple BacMam reagents can be readily used on the same cells. Recent improvements made to the BacMam system enable efficient transduction in a wider variety of cells, including neurons and neural stem cells (NSCs) with an easy, one-step protocol. To visualize autophagy, simply add the BacMam Autophagy Sensor to cells and incubate overnight for protein expression, then detect signal using standard GFP and/or RFP fluorescence settings on your instrument.

The LC3B protein plays a critical role in autophagy. Normally, this protein resides in the cytosol, but following cleavage and lipidation with phosphatidylethanolamine, LC3B associates with the phagophore. This localization can be used as a general marker for autophagic membranes. The p62 protein, also known as sequestosome (SQSTM1), is an ubitiquitin-binding protein that functions as a receptor for cargos destined to be degraded by the cellular autophagic machinery. When autophagy is induced, the p62 protein localizes to the autophagosomes and is subsequently degraded. Conversely, with the inhibition of autophagy, the p62 protein accumulates in the autophagosome. Thus, the subcellular localization of a p62-fluorescent protein chimera serves as a useful marker for the induction and inhibition of autophagy.

Additionally, by combining the acid-sensitive GFP (i.e., Emerald GFP) with an acid-insensitive RFP (i.e., TagRFP) in the tandem sensor RFP-GFP-LC3B, the change from an autophagosome (neutral pH) to the autolysosome (with an acidic pH) can be visualized by imaging the specific loss of the GFP fluorescence upon acidification of the autophagosome following lysosomal fusion. Upon induction of autophagy, the Premo Tandem Autophagy Sensor labels the punctate autophagosomes; these structures are positive for both GFP and RFP. Once the lysosome has fused, the pH drops, which quenches the GFP, making autolysosomes appear red. Additionally, combining the Premo Autophagy Tandem Sensor with the far-red emitting LysoTracker Deep Red (available separately) allows for a three-color analysis of autophagosomal/autolysomal/lysosomal dynamics.

Each Premo Autophagy assay kit includes chloroquine diphosphate, which is used as a negative control to inhibit autophagy.

Advantages of using Premo Autophagy assays
Multiplex inspired — combining the Premo Autophagy Tandem Sensor with the far-red emitting LysoTracker Deep Red allows for a three-color analysis of complete autophagic pathway dynamics
Highly efficient delivery system —>90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
Fast and convenient — simply add the Premo Autophagy reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
Robust — Premo Autophagy reagents are non-replicating in mammalian cells, lack observable cytopathic effect, and are suitable for biosafety level (BSL) 1 protocols
Multiple configurations: — Premo Autophagy sensors are available in the following chimeric configurations:
P36235: LC3B-GFP; control LC3B (G120A)-GFP; P36236: LC3B-RFP; control LC3B (G120A)-RFP; P36239: RFP-GFP-LC3B; P36240: GFP-p62; P36241: RFP-p62
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Compatible CellsMammalian Cells
DescriptionPremo Autophagy Sensor GFP-p62 Kit
Excitation/Emission488/506
For Use With (Equipment)Fluorescence Microscope
Kit ContentsAutophagy Sensor p62-GFP, and chloroquine diphosphate
Label TypeOther Label(s) or Dye(s)
Label or DyeGFP-p62
Product LinePremo
Product TypeAutophagy Sensor
Quantity1 kit
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 6°C, protected from light. Do Not Freeze.
Detection MethodFluorescence
FormatTube
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the Premo Autophagy Sensor products to study autophagy in cells in combination with a lentivirus transfection?

Lentivirus transfection may actually induce autophagy, skewing the results. Make certain to include appropriate controls.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3091973Certificate of AnalysisMar 30, 2025P36240
2971010Certificate of AnalysisOct 01, 2024P36239
2845301Certificate of AnalysisSep 19, 2024P36240
2976623Certificate of AnalysisAug 27, 2024P36241
2892688Certificate of AnalysisMay 21, 2024P36241
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (95)

Citations & References
Abstract
Degradation of heparan sulfate proteoglycans enhances oxidized-LDL-mediated autophagy and apoptosis in human endothelial cells.
Authors:Ding Z, Wang X, Khaidakov M, Liu S, Dai Y, Mehta JL,
Journal:Biochem Biophys Res Commun
PubMed ID:22910414
Cell surface heparan sulfate proteoglycans (HSPG) play an important role in atherogenesis. We hypothesized that degradation of HSPG may increase the binding of atherogenic oxidized low density lipoprotein (ox-LDL) to endothelial cells, and result in extensive HSPG degradation as well as autophagy and apoptosis. Primary human umbilical vein endothelial cells ... More
Pressure Overload-Induced Cardiac Dysfunction in Aged Male Adiponectin Knockout Mice Is Associated With Autophagy Deficiency.
Authors:Jahng JW, Turdi S, Kovacevic V, Dadson K, Li RK, Sweeney G,
Journal:
PubMed ID:25961840
'Heart failure is a leading cause of death, especially in the elderly or obese and diabetic populations. Various remodeling events have been characterized, which collectively contribute to the progression of heart failure. Of particular interest, autophagy has recently emerged as an important determinant of cardiac remodeling and function. Here, we ... More
MST1 functions as a key modulator of neurodegeneration in a mouse model of ALS.
Authors:Lee JK, Shin JH, Hwang SG, Gwag BJ, McKee AC, Lee J, Kowall NW, Ryu H, Lim DS, Choi EJ,
Journal:
PubMed ID:23818595
'Amyotrophic lateral sclerosis (ALS) is an adult-onset neurodegenerative disorder characterized by loss of motor neurons. Dominant mutations in the gene for superoxide dismutase 1 (SOD1) give rise to familial ALS by an unknown mechanism. Here we show that genetic deficiency of mammalian sterile 20-like kinase 1 (MST1) delays disease onset ... More
Apogossypol derivative BI-97C1 (Sabutoclax) targeting Mcl-1 sensitizes prostate cancer cells to mda-7/IL-24-mediated toxicity.
Authors:Dash R, Azab B, Quinn BA, Shen X, Wang XY, Das SK, Rahmani M, Wei J, Hedvat M, Dent P, Dmitriev IP, Curiel DT, Grant S, Wu B, Stebbins JL, Pellecchia M, Reed JC, Sarkar D, Fisher PB,
Journal:Proc Natl Acad Sci U S A
PubMed ID:21555592
'Limited options are available for treating patients with advanced prostate cancer (PC). Melanoma differentiation associated gene-7/interleukin-24 (mda-7/IL-24), an IL-10 family cytokine, exhibits pleiotropic anticancer activities without adversely affecting normal cells. We previously demonstrated that suppression of the prosurvival Bcl-2 family member, myeloid cell leukemia-1 (Mcl-1), is required for mda-7/IL-24-mediated apoptosis ... More
Nucleotide-oligomerizing domain-1 (NOD1) receptor activation induces pro-inflammatory responses and autophagy in human alveolar macrophages.
Authors:Juárez E, Carranza C, Hernández-Sánchez F, Loyola E, Escobedo D, León-Contreras JC, Hernández-Pando R, Torres M, Sada E,
Journal:
PubMed ID:25253572
'Nucleotide-binding oligomerizing domain-1 (NOD1) is a cytoplasmic receptor involved in recognizing bacterial peptidoglycan fragments that localize to the cytosol. NOD1 activation triggers inflammation, antimicrobial mechanisms and autophagy in both epithelial cells and murine macrophages. NOD1 mediates intracellular pathogen clearance in the lungs of mice; however, little is known about NOD1''s ... More
95 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection