Adapta™ Universal Kinase Assay Kit

Catalog number: PV5099
Have Questions?

Adapta™ Universal Kinase Assay Kit

Catalog number: PV5099
Have Questions?
Catalog Number
PV5099
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV50991 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Adapta™ Universal Kinase Assay Kit is a homogenous, fluorescence-based immunoassay for the detection of ADP produced by kinases or other ATP dependent enzymes.

For more information, please visit www.invitrogen.com/adapta.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Universal Kinase Assay
Assay Entry
Biochemical ADP accumulation
Detection Method
Fluorescent
Format
384-well Plate
Type
Kinase Assay Kit
Excitation/Emission
340⁄615, 665
For Use With (Application)
TR-FRET
Gene ID (Entrez)
1022, 1147, 120892, 1612, 3376, 5289, 5290, 5294, 8536
Label or Dye
Alexa Fluor™ 647, Europium
Product Line
Adapta™
Readout
End Point
Shipping Condition
Dry Ice
Target Entry
CAMK1, PIK3C2B, PIK3C3, PIK3CG, SPHK2, PI4KB, PIK3C2A, GSG2, LRRK2, CDK7/cyclin H/MNAT1, PI4KA, LRRK2 G2019S, CDK9/Cyclin T1, PIK3CA/PIK3R1, SPHK1, PIK3CD/PIK3R1, DAPK1
Target Enzyme
Kinases (Multiple)
Validated Kinases
CAMK1, CDK7, CDK9, CHUK, LRRK2, PI4KB, PIK3C2B, PIK3CA, PIK3CB, PIK3CD, PIK3CG, PIK3C2A

Contents & Storage

Adapta™ Universal Kinase Assay Kits are shipped on dry ice, and should be stored at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text