Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay | RNA Quantification, extended range |
Detection Method | Fluorescence |
For Use With (Equipment) | Qubit 4 and Qubit Flex Fluorometer |
No. of Reactions | 500 Reactions |
Quantitation Range | 0.1 to 20 μg |
Product Line | Qubit |
Shipping Condition | Dry Ice |
Unit Size | 500 assays |
Assay | RNA Quantification, broad range |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 100 Reactions |
Quantitation Range | 10 to 1200 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 100 assays |
Assay | RNA Quantification, broad range |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 500 Reactions |
Quantitation Range | 10 to 1200 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 500 assays |
Assay | RNA Quantification, extended range |
Detection Method | Fluorescence |
For Use With (Equipment) | Qubit 4 and Qubit Flex Fluorometer |
No. of Reactions | 100 Reactions |
Quantitation Range | 0.1 to 20 μg |
Product Line | Qubit |
Shipping Condition | Dry Ice |
Unit Size | 100 assays |
Assay | RNA Quantification, high sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 100 Reactions |
Quantitation Range | 4 to 200 ng |
Product Line | Quant-iT, Qubit |
Shipping Condition | Room Temperature |
Unit Size | 100 assays |
Assay | RNA Quantification, high sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Qubit Fluorometer |
No. of Reactions | 500 Reactions |
Quantitation Range | 4 to 200 ng |
Product Line | Quant-iT, Qubit |
Shipping Condition | Room Temperature |
Unit Size | 500 assays |
Catalog Number | Specifications | Unit Size | Assay | Quantitation Range | No. of Reactions | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q33224 | 500 assays | RNA Quantification, extended range | 0.1 to 20 μg | 500 Reactions | 369.65 Online price 386.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q10210 | 100 assays | RNA Quantification, broad range | 10 to 1200 ng | 100 Reactions | 123.65 Online price 129.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q10211 | 500 assays | RNA Quantification, broad range | 10 to 1200 ng | 500 Reactions | 369.65 Online price 384.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33223 | 100 assays | RNA Quantification, extended range | 0.1 to 20 μg | 100 Reactions | 123.65 Online price 129.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32852 | 100 assays | RNA Quantification, high sensitivity | 4 to 200 ng | 100 Reactions | 123.65 Online price 129.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32855 | 500 assays | RNA Quantification, high sensitivity | 4 to 200 ng | 500 Reactions | 369.65 Online price 383.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|