Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Invitrogen™

Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

Kits enable quick and sensitive detection of low and high abundance RNA, and can distinguish between RNA and DNA, protein, and free nucleotides.
Promo ImagePromotion
Promo code:P5780456
Gene Expression Promo
From RNA Isolation kits, reverse transcriptases to analysis of your gene with qPCR - upto 35% on all Gene Expression workflow productsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityAssay
Q33223Promo Image100 assaysRNA Quantification, ext range
Q10210Promo Image100 assaysRNA Quantification, BR
Q10211Promo Image500 assaysRNA Quantification, BR
Q33224Promo Image500 assaysRNA Quantification, ext range
Q32852Promo Image100 assaysRNA Quantification, HS
Q32855Promo Image500 assaysRNA Quantification, HS
Catalog number Q33223
Price (USD)
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Each
In stock
Add to cart
Quantity:
100 assays
Assay:
RNA Quantification, ext range
Recurring order eligible. Learn more »
Price (USD)
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Each
Add to cart
Ask our AI about this Product
Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.
Qubit RNA HS, BR, and XR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for RNA over DNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.

Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.

Qubit RNA XR Assay Kit

The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.

Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayRNA Quantification, ext range
For Use With (Equipment)Qubit 4 and Qubit Flex Fluorometer
No. of Reactions100 Reactions
Product LineQubit
Quantitation Range0.1 to 20 μg
Quantity100 assays
Shipping ConditionDry Ice
Detection MethodFluorescence
Unit SizeEach

Enhance your Qubit Fluorometer with the latest firmware

New Qubit 4 and Qubit Flex firmware features include importing sample IDs, renaming files before exporting, a countdown timer, and on-board troubleshooting guidance. Get the most out of your Qubit assays with these powerful new tools.

Download the latest Qubit firmware ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3192598Certificate of AnalysisJun 29, 2025Q33224
3148365Certificate of AnalysisJun 29, 2025Q10210
3244554Certificate of AnalysisJun 29, 2025Q33224
3148333Certificate of AnalysisJun 29, 2025Q32855
3223934Certificate of AnalysisJun 29, 2025Q10211
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (34)

Citations & References
Abstract
Expression Analysis on Archival Material Revisited: Isolation and Quantification of RNA Extracted From FFPE Samples.
Authors:Deben C, Zwaenepoel K, Boeckx C, Wouters A, Pauwels P, Peeters M, Lardon F, Baay M, Deschoolmeester V,
Journal:Diagn Mol Pathol
PubMed ID:23370428
'BACKGROUND:: Formalin-fixed paraffin-embedded (FFPE) tissue is the most readily available source of RNA for the gene expression studies. The main disadvantage is the poor quality of isolated RNA. Our group recently compared 5 commercially available RNA isolation kits and concluded that the RNeasy FFPE kit from Qiagen was the most ... More
Systems-Level Analysis of Nitrogen Starvation-Induced Modifications of Carbon Metabolism in a Chlamydomonas reinhardtii Starchless Mutant.
Authors:Blaby IK, Glaesener AG, Mettler T, Fitz-Gibbon ST, Gallaher SD, Liu B, Boyle NR, Kropat J, Stitt M, Johnson S, Benning C, Pellegrini M, Casero D, Merchant SS,
Journal:
PubMed ID:24280389
'To understand the molecular basis underlying increased triacylglycerol (TAG) accumulation in starchless (sta) Chlamydomonas reinhardtii mutants, we undertook comparative time-course transcriptomics of strains CC-4348 (sta6 mutant), CC-4349, a cell wall-deficient (cw) strain purported to represent the parental STA6 strain, and three independent STA6 strains generated by complementation of sta6 (CC-4565/STA6-C2, ... More
Analysis of in vivo expressed genes in Mannheimia haemolytica A1.
Authors:Lo RY, Sathiamoorthy S, Shewen PE
Journal:FEMS Microbiol Lett
PubMed ID:16984402
'The expression of Mannheimia haemolytica A1 genes during in vivo growth was examined by reverse transcriptase-polymerase chain reaction (RT-PCR) using total RNA extracted directly from M. haemolytica A1 recovered from pneumonic lungs of cattle. Primers specific for three groups of genes were used. Group 1 includes virulence-related genes: lktC, tbpB, ... More
High transcript level of fatty acid-binding protein 11 but not of very low-density lipoprotein receptor is correlated to ovarian follicle atresia in a teleost fish (Solea senegalensis).
Authors:Agulleiro MJ, André M, Morais S, Cerdà J, Babin PJ,
Journal:Biol Reprod
PubMed ID:17554079
'Transcripts encoding a fatty acid-binding protein (FABP), Fabp11, and two isoforms of very low-density lipoprotein receptor (Vldlr; vitellogenin receptor) were characterized from the ovary of Senegalese sole (Solea senegalensis). Phylogenetic analyses of vertebrate FABPs demonstrated that Senegalese sole Fabp11, as zebrafish (Danio rerio) homologous sequences, is part of a newly ... More
Comparative expression analysis of rpf-like genes of Mycobacterium tuberculosis H37Rv under different physiological stress and growth conditions.
Authors:Gupta RK, Srivastava BS, Srivastava R,
Journal:Microbiology
PubMed ID:20522500
'Mycobacterium tuberculosis H37Rv possesses five resuscitation-promoting factors, RpfA-E, which are required for the resuscitation of dormancy in mycobacteria induced by prolonged incubation of the culture in stationary phase. This study explores the transcriptional profile of all the rpf-like genes of M. tuberculosis H37Rv in the exponential phase, stationary phase, non-culturable ... More
34 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection