Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Control Slide along with PathoDx Respiratory Virus Panel (RVP) kit while performing direct immunofluorescence tests. Viral respiratory diseases in children and adults cause significant morbidity and mortality, especially in young children and older adults. Respiratory specimens (nasopharyngeal aspirates or swabs) are tested for the presence of viral antigen. Culture isolation/confirmation is the standard method for most clinical virology laboratories.
Description | PathoDx™ Respiratory Virus Panel Control Slides |
Type | Control |
Quantity | 5 Slides/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R62414 | Each | 5 Slides/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Negative Control Reagent contains fluorescein-labeled murine antibodies which do not react with the viral agents.
Negative Control Reagent (Red Cap) is one dropper bottle containing 10.0 mL of fluorescein-labeled purified murine IgG antibodies that are non-reactive with any respiratory virus, and Evans blue counterstain in a protein-stabilized buffer solution, with 0.098% sodium azide as preservative.
Each control slide has eight wells and each well is clearly marked with the type of virus-infected cell that is on the slide. One of the wells contains uninfected LLC-MK2 cells and the slide is fixed in acetone.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.