Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60839. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Plays a role in primary ciliogenesis by modulating actin polymerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliogenesis-associated TTC17-interacting protein
Gene Aliases: C2orf62; CATIP
UniProt ID: (Human) Q7Z7H3
Entrez Gene ID: (Human) 375307
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support