Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO

Catalog number: S7567
Have Questions?
Invitrogen™

SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO

Catalog number: S7567
Have Questions?
Catalog Number
S7567
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 x 50 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S75671 mL
-
S7563500 µL
-
S758520 x 50 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SYBR™ Green I Nucleic Acid Gel Stain is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels. Because SYBR™ Green I has greater sensitivity for dsDNA, it is especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR™ Green I stain is ideal for use with laser scanners. SYBR™ Green I stain provides:

Sensitivity—at least four times greater than ethidium bromide for DNA in agarose gels
Ease of use—gels soaked in diluted stain can be visualized without desalting
Compatibility—with UV transilluminators, gel documentation systems, and laser scanners
Flexibility—for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection

Product use
One mL stains 100 minigels. Various pack sizes are available.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescence
Label or Dye
SYBR Green I
Product Type
Nucleic Acid Gel Stain
Quantity
1 mL
Shipping Condition
Room Temperature
Target Molecule
DNA

Contents & Storage

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text