See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Electron Microscopes
Electron Microscope Accessories
Electron Microscope Specimen Mounts and Holders
Sample Holder Inserts
Thermo Scientific™
Sample Holder Inserts
Catalog number:
SAMPLEHOLDERINSERT
Related applications:
Materials Science
Have Questions?
Contact Us
Thermo Scientific™
Sample Holder Inserts
Catalog number:
SAMPLEHOLDERINSERT
Related applications:
Materials Science
Have Questions?
Contact Us
Cross-sectional imaging of coatings and multilayer samples becomes fast and easy with this metallurgical insert. Its smart, split clamping mechanism eliminates the need for screws and tools to clamp the sample.
Catalog Number
SAMPLEHOLDERINSERT
Unit Size
Each
Price (USD)
Request A Quote
Full specifications
Unit Size
Each
Save to list
Showing 1 of 1
Catalog Number
Specifications
Unit Size
Price (USD)
SAMPLEHOLDERINSERT
Full specifications
Each
Request A Quote
Unit Size
Each
Showing 1 of 1
Save to list
Key Specifications
Easy clamping without the need for screws or extra tools
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate