Transferrin From Human Serum, Fluorescein Conjugate
Transferrin From Human Serum, Fluorescein Conjugate
Invitrogen™

Transferrin From Human Serum, Fluorescein Conjugate

Transferrin is a monomeric serum glycoprotein (∼80,000 daltons) that binds to a specific receptor on the surface of vertebrate cellsRead more
Have Questions?
Catalog NumberQuantity
T28715 mg
Catalog number T2871
Price (USD)
490.00
5 mg
-
Add to cart
Quantity:
5 mg
Recurring order eligible. Learn more »
Price (USD)
490.00
5 mg
Add to cart
Ask our AI about this Product
Transferrin is a monomeric serum glycoprotein (∼80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis—our labeled LDL complexes are useful tools for studying this phenomenon. Once iron-carrying transferrin proteins are inside endosomes, the acidic environment favors dissociation of iron from the transferrin–receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane, where it is released from its receptor to scavenge more iron. Fluorescent transferrin conjugates can therefore be used with fluorescent LDL to distinguish the lysosomally directed and recycling endosomal pathways.

These experiments are typically performed by adding fluorescently labeled transferrin to cultured cells and analyzing them by microscopy. We offer a biotinylated transferrin conjugate and more than 10 fluorescent versions.

Transferrin Specifications:

Label (Ex/Em): Fluorescein (495/518)

Amount: 15 mg solid (contains 5 mg of transferrin conjugate)

Key Applications of Labeled Transferrin
Some of the many applications for labeled transferrin include:
• Image transferrin receptor dynamics using FRET
• Observe receptor trafficking in live cells by confocal laser-scanning microscopy
• Investigate events occurring during endosomal acidification
• Measure transferrin receptor binding affinity in mammals and parasites

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeClassic Dyes
Excitation/Emission495/518
FormSolid
Protein FamilyTransferrin
Quantity5 mg
Shipping ConditionRoom Temperature
Product TypeTransferrin
pH7.2
Unit Size5 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2855954Certificate of AnalysisMar 13, 2024T2871
2831351Certificate of AnalysisDec 14, 2023T2871
2561917Certificate of AnalysisJul 01, 2023T2871
2354386Certificate of AnalysisJun 16, 2021T2871
2105725Certificate of AnalysisJun 01, 2019T2871
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (103)

Citations & References
Abstract
Characterization of Endophilin B1b, a brain-specific membrane-associated lysophosphatidic acid acyl transferase with properties distinct from endophilin A1.
Authors:Modregger J, Schmidt AA, Ritter B, Huttner WB, Plomann M
Journal:J Biol Chem
PubMed ID:12456676
We have characterized mammalian endophilin B1, a novel member of the endophilins and a representative of their B subgroup. The endophilins B show the same domain organization as the endophilins A, which contain an N-terminal domain responsible for lipid binding and lysophosphatidic acid acyl transferase activity, a central coiled-coil domain ... More
Interactions between Piccolo and the actin/dynamin-binding protein Abp1 link vesicle endocytosis to presynaptic active zones.
Authors:Fenster SD, Kessels MM, Qualmann B, Chung WJ, Nash J, Gundelfinger ED, Garner CC
Journal:J Biol Chem
PubMed ID:12654920
'Piccolo is a high molecular weight multi-domain protein shown to be a structural component of the presynaptic CAZ (cytoskeletal matrix assembled at active zones). These features indicate that Piccolo may act to scaffold proteins involved in synaptic vesicle endo- and exocytosis near their site of action. To test this hypothesis, ... More
Receptor-mediated endocytosis of transferrin and the uptake of fe in K562 cells: identification of a nonlysosomal acidic compartment.
Authors:van Renswoude J, Bridges KR, Harford JB, Klausner RD
Journal:Proc Natl Acad Sci U S A
PubMed ID:6292894
'At physiological temperature, the Fe-carrier transferrin is taken up by K562 human erythroleukemia cells through receptor-mediated endocytosis. Both ligand (now minus Fe) and receptor recycle back to the cell surface where the receptor is rapidly reutilized. After endocytosis, transferrin becomes transiently lodged within an acidic compartment inside the cell, as ... More
Defective acidification of intracellular organelles in cystic fibrosis.
Authors:Barasch J, Kiss B, Prince A, Saiman L, Gruenert D, al-Awqati Q
Journal:Nature
PubMed ID:1712081
'The phenotype of cystic fibrosis (CF) includes abnormalities in transepithelial transport of Cl- (refs 1-5), decreased sialylation and increased sulphation and fucosylation of glycoproteins, and lung colonization with Pseudomonas. It is not apparent how these abnormalities are interrelated, nor how they result from loss of function of the CF gene-encoded ... More
Imaging of receptor trafficking by using alpha-bungarotoxin-binding-site-tagged receptors.
Authors:Sekine-Aizawa Y, Huganir RL,
Journal:Proc Natl Acad Sci U S A
PubMed ID:15563595
'alpha-Amino-3-hydroxy-5-methyl-4-isoxazole-propionate (AMPA) receptors mediate excitatory synaptic transmission and are dynamically regulated during synaptic plasticity in the CNS. The membrane trafficking of AMPA receptors to synapses is critical for the regulation of the efficacy of excitatory synaptic transmission. Direct imaging of AMPA receptors in various cell compartments is important to dissecting ... More
103 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection