Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With | Siliconizing Glass Surfaces |
Standard Components | 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane |
Standard Concentration | 98 to 100% |
Item Description | 480 mL |
Quantity | 480 mL |
Storage | Room Temperature |
Product Type | Hydrocarbon Soluble Siliconizing Fluid |
Unit Size | Each |
For Use With | GC |
Hazards | Corrosive to Eyes/Skin/Respiratory Tract |
Standard Components | 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxane |
Item Description | 120 mL |
Quantity | 120 mL |
Product Type | Hydrocarbon Soluble Siliconizing Fluid |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
TS-42801 | Each | 480 mL | 810.25 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
TS-42800 | Each | 120 mL | 204.86 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Hydrocarbon Soluble Siliconizing Fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid.
The fluid is acidic only during application; after application, the surface is neutral.