Yeast Mitochondrial Stain Sampler Kit
Get the sharpest, brightest images. Try new Invitrogen™ SlowFade™ Glass antifade RIMS mountant with refractive index of 1.52. Learn more »
Yeast Mitochondrial Stain Sampler Kit
Invitrogen™

Yeast Mitochondrial Stain Sampler Kit

The Yeast Mitochondrial Stain Sampler Kit contains sample quantities of five different cell-permeant probes that have been found to labelRead more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Catalog NumberQuantity
Y7530Promo Image1 kit
Catalog number Y7530
Price (USD)
426.00
1 kit
Estimated availability date 25-Aug-2025
Add to cart
Quantity:
1 kit
Recurring order eligible. Learn more »
Price (USD)
426.00
1 kit
Add to cart
Ask our AI about this Product
The Yeast Mitochondrial Stain Sampler Kit contains sample quantities of five different cell-permeant probes that have been found to label mitochondria in live yeast. In addition to the well characterized mitochondrial stains rhodamine 123, and DiOC6(3), this kit includes rhodamine B hexyl ester, MitoTracker™ Green FM and SYTO™ 18 yeast mitochondrial stain, a proprietary green fluorescent nucleic acid stain. Each of these probes is sequestered in functioning mitochondria.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Detection MethodFluorescence
For Use With (Equipment)Fluorescence Microscope
Quantity1 kit
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeYeast Mitochondrial Stain Sampler Kit
SubCellular LocalizationMitochondria
Unit Size1 kit
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2633275Certificate of AnalysisAug 24, 2023Y7530
2507889Certificate of AnalysisSep 01, 2022Y7530
2381725Certificate of AnalysisDec 02, 2021Y7530
2261468Certificate of AnalysisAug 25, 2020Y7530
2165136Certificate of AnalysisJan 13, 2020Y7530
5 results displayed, search above for a specific certificate

Molecular Probes® Handbook

Citations & References (4)

Citations & References
Abstract
Transcriptional Profiling of Saccharomyces cerevisiae Reveals the Impact of Variation of a Single Transcription Factor on Differential Gene Expression in 4NQO, Fermentable, and Nonfermentable Carbon Sources.
Authors:
Journal:G3 (Bethesda)
PubMed ID:29208650
The effect of copy number on α-synuclein's toxicity and its protective role in Bax-induced apoptosis, in yeast.
Authors:
Journal:Biosci Rep
PubMed ID:32794578
The genetic basis of aneuploidy tolerance in wild yeast.
Authors:
Journal:Elife
PubMed ID:31909711
Apoptosis, Induced by Human α-Synuclein in Yeast, Can Occur Independent of Functional Mitochondria.
Authors:
Journal:Cells
PubMed ID:33003464
4 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection