Thermo Scientific™
SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Catalog number: 3751RC-HR
Have Questions?
Thermo Scientific™
SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Catalog number: 3751RC-HR
Have Questions?
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Catalog Number
3751RC-HR
Unit Size
Case of 4800
Model
200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)200 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.97 in.
Length (Metric)5.00 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeGreen
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 4800
Model
20 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)20 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model20 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.95 in.
Length (Metric)4.86 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeRed
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 4800
Model
20 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)20 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model20 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.95 in.
Length (Metric)4.86 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeRed
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 4800
Model
300 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)300 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model300 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)2.26 in.
Length (Metric)5.75 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeOrange
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 4800
Model
300 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)300 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model300 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)2.26 in.
Length (Metric)5.75 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeOrange
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 3072
Model
1200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)1200 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model1200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.42 in.
Length (Metric)8.67 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeYellow
Product LineSoftfit-L
Unit SizeCase of 3072
Catalog Number
Unit Size
Case of 3072
Model
1200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)1200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model1200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.42 in.
Length (Metric)8.67 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeYellow
Product LineSoftfit-L
Unit SizeCase of 3072
Catalog Number
Unit Size
Case of 4800
Model
200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.97 in.
Length (Metric)5.00 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeGreen
Product LineSoftfit-L
Unit SizeCase of 4800
Catalog Number
Unit Size
Case of 3072
Model
1000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model1000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.39 in.
Length (Metric)8.61 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeBlue
Product LineSoftfit-L
Unit SizeCase of 3072
Catalog Number
Unit Size
Case of 3072
Model
1000 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Price (ARS)
Full specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model1000 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.39 in.
Length (Metric)8.61 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeBlue
Product LineSoftfit-L
Unit SizeCase of 3072
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeModelPrice (ARS)
3751RC-HRFull specifications
Case of 4800200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)200 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.97 in.
Length (Metric)5.00 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeGreen
Product LineSoftfit-L
Unit SizeCase of 4800
3721RC-HRFull specifications
Case of 480020 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)20 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model20 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.95 in.
Length (Metric)4.86 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeRed
Product LineSoftfit-L
Unit SizeCase of 4800
3722RC-HRFull specifications
Case of 480020 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)20 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model20 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.95 in.
Length (Metric)4.86 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeRed
Product LineSoftfit-L
Unit SizeCase of 4800
3731RC-HRFull specifications
Case of 4800300 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)300 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model300 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)2.26 in.
Length (Metric)5.75 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeOrange
Product LineSoftfit-L
Unit SizeCase of 4800
3732RC-HRFull specifications
Case of 4800300 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)300 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model300 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)2.26 in.
Length (Metric)5.75 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeOrange
Product LineSoftfit-L
Unit SizeCase of 4800
3741RC-HRFull specifications
Case of 30721200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)1200 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model1200 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.42 in.
Length (Metric)8.67 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeYellow
Product LineSoftfit-L
Unit SizeCase of 3072
3742RC-HRFull specifications
Case of 30721200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)1200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model1200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.42 in.
Length (Metric)8.67 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeYellow
Product LineSoftfit-L
Unit SizeCase of 3072
3752RC-HRFull specifications
Case of 4800200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model200 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)1.97 in.
Length (Metric)5.00 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeGreen
Product LineSoftfit-L
Unit SizeCase of 4800
3781RC-HRFull specifications
Case of 30721000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)1000 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
AutoclavableNot Autoclavable
Model1000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.39 in.
Length (Metric)8.61 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeBlue
Product LineSoftfit-L
Unit SizeCase of 3072
3782RC-HRFull specifications
Case of 30721000 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle KitRequest A Quote
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
AutoclavableNot Autoclavable
Model1000 μL, Non-Filtered, Non-sterile, Hinged Rack, TipCycle Recycle Kit
Length (English)3.39 in.
Length (Metric)8.61 cm
Clean ClaimsDNase, RNase, DNA, Pryogen Free
FormatHinged Rack
Tip StyleSpecialty
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color CodeBlue
Product LineSoftfit-L
Unit SizeCase of 3072
Showing 10 of 10
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume