Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Search Thermo Fisher Scientific
Description | Amira Software for biomedical and life science research. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (ARS) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AMIRA | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
From imaging to understanding mechanisms of life, Amira Software supports entire discovery workflows. Process and visualize data from any imaging modality, at any scale, of any size. Perform routine and advanced analyses from a single, universal software application. Learn more about some of Amira’s use examples in: Cell biology Neuroscience and brain research Preclinical and clinical research imaging.