Thermo Scientific™

Transfer Assembly Jumper Tubing, TPE, Non-aseptic

Catalog number: SH31201.11
Thermo Scientific™

Transfer Assembly Jumper Tubing, TPE, Non-aseptic

Catalog number: SH31201.11
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
 
Catalog Number
SH31201.11
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialSilicone
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU Insert
Price (ARS)
Full specifications
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialSilicone
Unit Size1 each
Showing 10 of 90Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (ARS)
SH31201.11Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
SH31201.01Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
SH31205.11Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU InsertRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
SH31201.02Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
SH31205.12Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU InsertRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
SH31201.03Full specifications
1 eachTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
SH31205.13Full specifications
1 eachTransfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU InsertRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialC-Flex
Unit Size1 each
SH31205.01Full specifications
1 eachTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU InsertRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialSilicone
Unit Size1 each
SH31201.12Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clampRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp
Unit Size1 each
SH31205.02Full specifications
1 eachTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU InsertRequest A Quote
DescriptionTransfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU Insert
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesMPU Body and MPU Insert
Tubing MaterialSilicone
Unit Size1 each
Showing 10 of 90Show more
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.

• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance

Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.

All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.

These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text