| SH31489.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connector and 1 1/2 Tri-Clamp | Contact Us › |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Braided Silicone, AseptiQuik G Connector and 1 1/2 Tri-Clamp | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Unit Size | 1 each |
|
| SH31379.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik S Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31379.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik S Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31379.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/8 in. ID Silicone, AseptiQuik S Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik S Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31197.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik G Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31197.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik G Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31197.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik G Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31380.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik G Connector and 3/4 Tri-Clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik G Connector and 3/4 Tri-Clamp | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik G Connector and 3/4 Tri-Clamp | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31379.11 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik S Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID Silicone, AseptiQuik S Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik S Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31379.12 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, AseptiQuik S Connectors | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID Silicone, AseptiQuik S Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | AseptiQuik S Connectors | Tubing Material | Silicone | Unit Size | 1 each |
|