Megestrol acetate
Thermo Scientific Chemicals

Megestrol acetate

Have Questions?
Catalog NumberQuantity
4657900505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465790050
Price (EUR)
160,65
Online Exclusive
189,00
Save 28,35 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
160,65
Online Exclusive
189,00
Save 28,35 (15%)
Each
Add to cart
Chemical Identifiers
CAS595-33-5
IUPAC Name(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate
Molecular FormulaC24H32O4
InChI KeyRQZAXGRLVPAYTJ-GQFGMJRRSA-N
SMILESCC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point213°C to 220°C
HPLC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0444585Certificate of AnalysisFeb 03, 2025465790010, 465790050
A0420390Certificate of AnalysisJan 06, 2023465790010, 465790050
A0444585Certificate of AnalysisAug 09, 2022465790010, 465790050
3 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Carcinogenicity
Category 2
Specific target organ toxicity - (repeated exposure)
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H351 - Suspected of causing cancer

H373 - May cause damage to organs through prolonged or repeated exposure

Precautionary Statements

P201 - Obtain special instructions before use

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P308 + P313 - IF exposed or concerned: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection