Abiraterone acetate
Thermo Scientific Chemicals

Abiraterone acetate

Have Questions?
Catalog NumberQuantity
466732500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466732500
Price (EUR)
72,25
Online Exclusive
85,00
Save 12,75 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
72,25
Online Exclusive
85,00
Save 12,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS154229-18-2
IUPAC Name(7S,9aR,11aS)-9a,11a-dimethyl-1-(pyridin-3-yl)-3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Molecular FormulaC26H33NO2
InChI KeyUVIQSJCZCSLXRZ-HMMZIKKISA-N
SMILESCC(=O)O[C@H]1CC[C@]2(C)C3CC[C@@]4(C)C(CC=C4C4=CC=CN=C4)C3CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)crystalline powder
HPLC>=98.0 %
Melting point143.0°C to 147.0°C
Appearance (Color)White to almost white
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471799Certificate of AnalysisMay 17, 2025466732500, 466730010
A0465356Certificate of AnalysisAug 21, 2024466732500, 466730010
A0453085Certificate of AnalysisAug 25, 2023466732500, 466730010
A0450193Certificate of AnalysisMay 02, 2023466732500, 466730010
A0418873Certificate of AnalysisDec 05, 2022466732500, 466730010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Reproductive Toxicity
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H361 - Suspected of damaging fertility or the unborn child

Precautionary Statements

P201 - Obtain special instructions before use

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection