Desonide
Thermo Scientific Chemicals

Desonide

Have Questions?
Catalog NumberQuantity
467452500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 467452500
Price (EUR)
26,18
Online Exclusive
30,80
Save 4,62 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
26,18
Online Exclusive
30,80
Save 4,62 (15%)
Each
Add to cart
Chemical Identifiers
CAS638-94-8
IUPAC Name(2S,4R,8S,12S)-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Molecular FormulaC24H32O6
InChI KeyWBGKWQHBNHJJPZ-YWZQBGSISA-N
SMILESCC1(C)O[C@@H]2C[C@H]3C4CCC5=CC(=O)C=CC5(C)[C@H]4C(O)CC3(C)[C@@]2(O1)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.5 % (1 g, 105°C, 3 hr)
Appearance (Color)White to almost white
HPLC>=97.5 %
Specific optical rotation+104° to +110° (20°C, 589 nm) (10 mg/ml in dioxane)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0419851Certificate of AnalysisJan 05, 2023467452500, 467450050, 467450010
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection